Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197092887-197093146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPM
Entrez Gene ID
259266
Gene Name
abnormal spindle microtubule assembly
Gene Aliases
ASP, Calmbp1, MCPH5
UniGene
Hs.121028
Gene Chromosome Location
Chr.1: 197084127-197146694 on Build GRCh38
Transcripts
NM_001206846.1 NM_018136.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901653 c.9286C>T p.R3096* Chr.1: 197093060-197093060 on Build GRCh38
5832442 c.9246_9247insT p.T3083fs*41 Chr.1: 197093099-197093100 on Build GRCh38
6060742 c.9237T>A p.F3079L Chr.1: 197093109-197093109 on Build GRCh38
1243089 c.9254T>C p.I3085T Chr.1: 197093092-197093092 on Build GRCh38
6606035 c.9284T>C p.V3095A Chr.1: 197093062-197093062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758783903 Chr.1: 197093117-197093117 on Build GRCh38
rs369393071 Chr.1: 197093044-197093044 on Build GRCh38
rs779419715 Chr.1: 197093018-197093018 on Build GRCh38
rs138138436 Chr.1: 197093092-197093092 on Build GRCh38
rs947703198 Chr.1: 197092932-197092932 on Build GRCh38
rs772722368 Chr.1: 197093032-197093032 on Build GRCh38
rs151142538 Chr.1: 197093070-197093070 on Build GRCh38
rs146444278 Chr.1: 197093059-197093059 on Build GRCh38
rs147572084 Chr.1: 197092989-197092989 on Build GRCh38
rs749449214 Chr.1: 197093102-197093103 on Build GRCh38
rs767387811 Chr.1: 197093072-197093072 on Build GRCh38
rs1046101934 Chr.1: 197092936-197092936 on Build GRCh38
rs376811340 Chr.1: 197093034-197093034 on Build GRCh38
rs139097869 Chr.1: 197093096-197093096 on Build GRCh38
rs760152566 Chr.1: 197093075-197093075 on Build GRCh38
rs988431628 Chr.1: 197092998-197092998 on Build GRCh38
rs956946388 Chr.1: 197092990-197092990 on Build GRCh38
rs199503603 Chr.1: 197093050-197093050 on Build GRCh38
rs201050851 Chr.1: 197093100-197093100 on Build GRCh38
rs776753470 Chr.1: 197093045-197093045 on Build GRCh38
rs147005963 Chr.1: 197093074-197093074 on Build GRCh38
rs927829765 Chr.1: 197092951-197092951 on Build GRCh38
rs753507869 Chr.1: 197093011-197093011 on Build GRCh38
rs1032478911 Chr.1: 197092947-197092947 on Build GRCh38
rs756352237 Chr.1: 197093006-197093013 on Build GRCh38
rs36074161 Chr.1: 197093116-197093117 on Build GRCh38
rs34981455 Chr.1: 197093116-197093116 on Build GRCh38
rs774143329 Chr.1: 197093060-197093060 on Build GRCh38
rs185289429 Chr.1: 197092999-197092999 on Build GRCh38
rs916361125 Chr.1: 197092930-197092930 on Build GRCh38
rs535630513 Chr.1: 197093109-197093109 on Build GRCh38
rs377447809 Chr.1: 197093078-197093078 on Build GRCh38
rs1007218566 Chr.1: 197092920-197092920 on Build GRCh38
rs199422186 Chr.1: 197093108-197093108 on Build GRCh38
rs755435140 Chr.1: 197093014-197093014 on Build GRCh38
rs541141292 Chr.1: 197093087-197093087 on Build GRCh38
rs753505219 Chr.1: 197093086-197093086 on Build GRCh38
rs376736125 Chr.1: 197093003-197093003 on Build GRCh38
rs748727275 Chr.1: 197093026-197093026 on Build GRCh38
rs777826220 Chr.1: 197093045-197093048 on Build GRCh38

View Full Product Details