Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193135232-193135465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_024529.4 XM_006711537.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6917946 c.325G>T p.D109Y Chr.1: 193135408-193135408 on Build GRCh38
7448262 c.312A>G p.T104T Chr.1: 193135395-193135395 on Build GRCh38
331152 c.366_367TC>CT p.T122>? Chr.1: 193135449-193135450 on Build GRCh38
1337374 c.340T>G p.L114V Chr.1: 193135423-193135423 on Build GRCh38
5072072 c.364A>G p.T122A Chr.1: 193135447-193135447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754491728 Chr.1: 193135370-193135370 on Build GRCh38
rs546046048 Chr.1: 193135267-193135267 on Build GRCh38
rs747337987 Chr.1: 193135395-193135395 on Build GRCh38
rs753791966 Chr.1: 193135369-193135369 on Build GRCh38
rs767218233 Chr.1: 193135341-193135341 on Build GRCh38
rs778469738 Chr.1: 193135371-193135371 on Build GRCh38
rs761807789 Chr.1: 193135431-193135431 on Build GRCh38
rs1046974673 Chr.1: 193135290-193135290 on Build GRCh38
rs886041278 Chr.1: 193135438-193135438 on Build GRCh38
rs369254776 Chr.1: 193135399-193135399 on Build GRCh38
rs760287746 Chr.1: 193135390-193135390 on Build GRCh38
rs563211268 Chr.1: 193135275-193135275 on Build GRCh38
rs952579366 Chr.1: 193135417-193135417 on Build GRCh38
rs965169255 Chr.1: 193135373-193135373 on Build GRCh38
rs747397191 Chr.1: 193135376-193135376 on Build GRCh38
rs1016696958 Chr.1: 193135334-193135334 on Build GRCh38
rs769833729 Chr.1: 193135419-193135419 on Build GRCh38
rs752356591 Chr.1: 193135384-193135384 on Build GRCh38
rs771258964 Chr.1: 193135398-193135398 on Build GRCh38
rs201766013 Chr.1: 193135375-193135375 on Build GRCh38
rs886988441 Chr.1: 193135317-193135317 on Build GRCh38
rs766379008 Chr.1: 193135368-193135368 on Build GRCh38
rs868045207 Chr.1: 193135421-193135421 on Build GRCh38
rs374834265 Chr.1: 193135416-193135416 on Build GRCh38
rs750182992 Chr.1: 193135345-193135345 on Build GRCh38
rs760710336 Chr.1: 193135353-193135353 on Build GRCh38
rs1005361595 Chr.1: 193135328-193135328 on Build GRCh38
rs75227966 Chr.1: 193135385-193135385 on Build GRCh38
rs999173509 Chr.1: 193135337-193135337 on Build GRCh38
rs778135669 Chr.1: 193135388-193135388 on Build GRCh38

View Full Product Details