Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8823135-8823406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
UniGene
Hs.620532
Gene Chromosome Location
Chr.12: 8822472-8887202 on Build GRCh38
Transcripts
NM_144670.5 XM_011520566.2 XM_011520567.2 XM_017018868.1 XM_017018869.1 XM_017018870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
431955 c.105C>A p.S35S Chr.12: 8823224-8823224 on Build GRCh38
6531410 c.151A>C p.K51Q Chr.12: 8823270-8823270 on Build GRCh38
695501 c.72G>T p.L24L Chr.12: 8823191-8823191 on Build GRCh38
1243826 c.177G>T p.K59N Chr.12: 8823296-8823296 on Build GRCh38
6138329 c.104C>T p.S35F Chr.12: 8823223-8823223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755096284 Chr.12: 8823343-8823343 on Build GRCh38
rs769565295 Chr.12: 8823373-8823373 on Build GRCh38
rs772906952 Chr.12: 8823192-8823192 on Build GRCh38
rs751739482 Chr.12: 8823341-8823341 on Build GRCh38
rs779067965 Chr.12: 8823241-8823241 on Build GRCh38
rs781346154 Chr.12: 8823350-8823350 on Build GRCh38
rs745711489 Chr.12: 8823292-8823293 on Build GRCh38
rs758676296 Chr.12: 8823338-8823338 on Build GRCh38
rs77657214 Chr.12: 8823340-8823340 on Build GRCh38
rs370012102 Chr.12: 8823382-8823382 on Build GRCh38
rs1036875344 Chr.12: 8823222-8823222 on Build GRCh38
rs771893464 Chr.12: 8823371-8823372 on Build GRCh38
rs79149293 Chr.12: 8823277-8823277 on Build GRCh38
rs770708682 Chr.12: 8823164-8823166 on Build GRCh38
rs989735586 Chr.12: 8823270-8823270 on Build GRCh38
rs557136055 Chr.12: 8823344-8823344 on Build GRCh38
rs202006422 Chr.12: 8823205-8823205 on Build GRCh38
rs374395580 Chr.12: 8823258-8823258 on Build GRCh38
rs1051881399 Chr.12: 8823246-8823246 on Build GRCh38
rs371564965 Chr.12: 8823320-8823320 on Build GRCh38
rs939451589 Chr.12: 8823290-8823290 on Build GRCh38
rs757200771 Chr.12: 8823256-8823256 on Build GRCh38
rs764547962 Chr.12: 8823209-8823209 on Build GRCh38
rs770444077 Chr.12: 8823285-8823285 on Build GRCh38
rs886042937 Chr.12: 8823294-8823295 on Build GRCh38
rs959789272 Chr.12: 8823172-8823172 on Build GRCh38
rs772183903 Chr.12: 8823165-8823165 on Build GRCh38
rs781254832 Chr.12: 8823263-8823263 on Build GRCh38
rs765032988 Chr.12: 8823190-8823190 on Build GRCh38
rs1057517901 Chr.12: 8823290-8823290 on Build GRCh38
rs754247426 Chr.12: 8823218-8823218 on Build GRCh38
rs747225412 Chr.12: 8823335-8823335 on Build GRCh38
rs767370461 Chr.12: 8823278-8823278 on Build GRCh38
rs750524549 Chr.12: 8823187-8823187 on Build GRCh38
rs367999763 Chr.12: 8823261-8823261 on Build GRCh38
rs746978816 Chr.12: 8823169-8823169 on Build GRCh38
rs774360683 Chr.12: 8823247-8823247 on Build GRCh38
rs17792974 Chr.12: 8823305-8823305 on Build GRCh38
rs185519272 Chr.12: 8823315-8823315 on Build GRCh38
rs1031874914 Chr.12: 8823293-8823293 on Build GRCh38
rs756640163 Chr.12: 8823208-8823208 on Build GRCh38
rs193079087 Chr.12: 8823207-8823207 on Build GRCh38
rs200007710 Chr.12: 8823178-8823178 on Build GRCh38
rs201185025 Chr.12: 8823224-8823224 on Build GRCh38
rs909507521 Chr.12: 8823384-8823384 on Build GRCh38
rs915537479 Chr.12: 8823272-8823272 on Build GRCh38
rs764639990 Chr.12: 8823311-8823311 on Build GRCh38
rs778669722 Chr.12: 8823260-8823260 on Build GRCh38
rs774086428 Chr.12: 8823229-8823231 on Build GRCh38
rs370199872 Chr.12: 8823168-8823168 on Build GRCh38
rs773944991 Chr.12: 8823298-8823298 on Build GRCh38
rs761671142 Chr.12: 8823188-8823188 on Build GRCh38
rs999028234 Chr.12: 8823283-8823283 on Build GRCh38
rs933811492 Chr.12: 8823237-8823237 on Build GRCh38
rs190479708 Chr.12: 8823319-8823319 on Build GRCh38
rs79315547 Chr.12: 8823359-8823359 on Build GRCh38

View Full Product Details