Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82011502-82011769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
UniGene
Hs.298351
Gene Chromosome Location
Chr.17: 81977550-82018929 on Build GRCh38
Transcripts
NM_001251888.1 NM_024083.3 NR_045351.1 XM_011523601.2 XM_011523602.2 XM_017025036.1 XM_017025037.1 XM_017025038.1 XM_017025039.1 XM_017025040.1 XM_017025041.1 XM_017025042.1 XM_017025043.1 XM_017025044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2805901 c.1300+4C>T p.? Chr.17: 82011609-82011609 on Build GRCh38
6433573 c.1276A>G p.N426D Chr.17: 82011581-82011581 on Build GRCh38
4747546 c.1269_1300+13del45 p.? Chr.17: 82011574-82011618 on Build GRCh38
4130835 c.1260G>C p.V420V Chr.17: 82011565-82011565 on Build GRCh38
4273373 c.1069+4C>T p.? Chr.17: 82011609-82011609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770644643 Chr.17: 82011569-82011569 on Build GRCh38
rs774273592 Chr.17: 82011651-82011651 on Build GRCh38
rs368381332 Chr.17: 82011646-82011646 on Build GRCh38
rs145237583 Chr.17: 82011657-82011657 on Build GRCh38
rs941972008 Chr.17: 82011663-82011663 on Build GRCh38
rs199630997 Chr.17: 82011545-82011545 on Build GRCh38
rs373635581 Chr.17: 82011571-82011571 on Build GRCh38
rs750787852 Chr.17: 82011610-82011610 on Build GRCh38
rs763059544 Chr.17: 82011530-82011530 on Build GRCh38
rs535173971 Chr.17: 82011650-82011650 on Build GRCh38
rs759454582 Chr.17: 82011732-82011732 on Build GRCh38
rs775524594 Chr.17: 82011524-82011524 on Build GRCh38
rs746572416 Chr.17: 82011562-82011562 on Build GRCh38
rs756835393 Chr.17: 82011632-82011632 on Build GRCh38
rs896085963 Chr.17: 82011542-82011542 on Build GRCh38
rs889280525 Chr.17: 82011692-82011692 on Build GRCh38
rs370380394 Chr.17: 82011534-82011534 on Build GRCh38
rs370514086 Chr.17: 82011587-82011587 on Build GRCh38
rs377597070 Chr.17: 82011586-82011586 on Build GRCh38
rs774003805 Chr.17: 82011676-82011676 on Build GRCh38
rs927388499 Chr.17: 82011698-82011698 on Build GRCh38
rs762736907 Chr.17: 82011604-82011604 on Build GRCh38
rs745656872 Chr.17: 82011641-82011641 on Build GRCh38
rs1035958797 Chr.17: 82011665-82011665 on Build GRCh38
rs780936267 Chr.17: 82011639-82011639 on Build GRCh38
rs1032512067 Chr.17: 82011743-82011743 on Build GRCh38
rs751786019 Chr.17: 82011531-82011531 on Build GRCh38
rs553844173 Chr.17: 82011544-82011544 on Build GRCh38
rs1005121420 Chr.17: 82011563-82011563 on Build GRCh38
rs201221183 Chr.17: 82011647-82011647 on Build GRCh38
rs758044279 Chr.17: 82011548-82011548 on Build GRCh38
rs897526318 Chr.17: 82011672-82011672 on Build GRCh38
rs937330911 Chr.17: 82011731-82011731 on Build GRCh38
rs761893252 Chr.17: 82011588-82011588 on Build GRCh38
rs189420247 Chr.17: 82011649-82011649 on Build GRCh38
rs763006339 Chr.17: 82011584-82011584 on Build GRCh38
rs755510254 Chr.17: 82011620-82011620 on Build GRCh38
rs751099966 Chr.17: 82011629-82011629 on Build GRCh38
rs180924216 Chr.17: 82011721-82011721 on Build GRCh38
rs775471582 Chr.17: 82011583-82011583 on Build GRCh38
rs578021575 Chr.17: 82011691-82011691 on Build GRCh38
rs373943690 Chr.17: 82011617-82011617 on Build GRCh38
rs80039090 Chr.17: 82011737-82011737 on Build GRCh38
rs149012488 Chr.17: 82011555-82011555 on Build GRCh38
rs756669382 Chr.17: 82011537-82011537 on Build GRCh38
rs374992477 Chr.17: 82011621-82011621 on Build GRCh38
rs754342953 Chr.17: 82011619-82011619 on Build GRCh38
rs745709769 Chr.17: 82011575-82011575 on Build GRCh38
rs766406184 Chr.17: 82011609-82011609 on Build GRCh38

View Full Product Details