Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147866677-147866926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP10
Entrez Gene ID
79658
Gene Name
Rho GTPase activating protein 10
Gene Aliases
GRAF2, PS-GAP, PSGAP
UniGene
Hs.368631
Gene Chromosome Location
Chr.4: 147732063-148072781 on Build GRCh38
Transcripts
NM_024605.3 XM_005263215.3 XM_017008602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051869 c.646G>T p.E216* Chr.4: 147866760-147866760 on Build GRCh38
7103498 c.630C>G p.F210L Chr.4: 147866744-147866744 on Build GRCh38
3127221 c.607delT p.Q205fs*46 Chr.4: 147866721-147866721 on Build GRCh38
7238687 c.616G>T p.G206W Chr.4: 147866730-147866730 on Build GRCh38
357604 c.630C>T p.F210F Chr.4: 147866744-147866744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143552684 Chr.4: 147866732-147866732 on Build GRCh38
rs879247986 Chr.4: 147866746-147866748 on Build GRCh38
rs773514097 Chr.4: 147866786-147866786 on Build GRCh38
rs997390021 Chr.4: 147866896-147866896 on Build GRCh38
rs145720035 Chr.4: 147866779-147866779 on Build GRCh38
rs771467245 Chr.4: 147866803-147866803 on Build GRCh38
rs200992418 Chr.4: 147866832-147866832 on Build GRCh38
rs760894570 Chr.4: 147866825-147866825 on Build GRCh38
rs775580937 Chr.4: 147866821-147866821 on Build GRCh38
rs765671085 Chr.4: 147866749-147866749 on Build GRCh38
rs774899236 Chr.4: 147866808-147866808 on Build GRCh38
rs751506588 Chr.4: 147866846-147866846 on Build GRCh38
rs4463051 Chr.4: 147866772-147866772 on Build GRCh38
rs746544060 Chr.4: 147866752-147866752 on Build GRCh38
rs752372593 Chr.4: 147866810-147866810 on Build GRCh38
rs779745174 Chr.4: 147866746-147866746 on Build GRCh38
rs966499839 Chr.4: 147866874-147866874 on Build GRCh38
rs199963512 Chr.4: 147866754-147866754 on Build GRCh38
rs1010224811 Chr.4: 147866759-147866759 on Build GRCh38
rs750401483 Chr.4: 147866719-147866719 on Build GRCh38
rs767630556 Chr.4: 147866819-147866819 on Build GRCh38
rs766477897 Chr.4: 147866844-147866844 on Build GRCh38
rs143731526 Chr.4: 147866791-147866791 on Build GRCh38
rs375524610 Chr.4: 147866826-147866826 on Build GRCh38
rs574955455 Chr.4: 147866724-147866724 on Build GRCh38
rs747766567 Chr.4: 147866774-147866774 on Build GRCh38
rs759592358 Chr.4: 147866817-147866817 on Build GRCh38
rs923860706 Chr.4: 147866862-147866862 on Build GRCh38
rs752311692 Chr.4: 147866868-147866868 on Build GRCh38
rs892746595 Chr.4: 147866757-147866757 on Build GRCh38
rs754473364 Chr.4: 147866787-147866787 on Build GRCh38
rs377701328 Chr.4: 147866867-147866867 on Build GRCh38
rs368488295 Chr.4: 147866827-147866827 on Build GRCh38
rs754446745 Chr.4: 147866849-147866849 on Build GRCh38
rs771061630 Chr.4: 147866828-147866855 on Build GRCh38
rs995191186 Chr.4: 147866768-147866768 on Build GRCh38
rs778679784 Chr.4: 147866833-147866833 on Build GRCh38
rs765059491 Chr.4: 147866711-147866711 on Build GRCh38

View Full Product Details