Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 48941114-48941339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF2
Entrez Gene ID
10564
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 2
Gene Aliases
BIG2, PVNH2, dJ1164I10.1
UniGene
Hs.62578
Gene Chromosome Location
Chr.20: 48921721-49036693 on Build GRCh38
Transcripts
NM_006420.2 XM_005260252.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4803060 c.140T>G p.I47R Chr.20: 48941217-48941217 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931213978 Chr.20: 48941205-48941205 on Build GRCh38
rs1015232199 Chr.20: 48941308-48941308 on Build GRCh38
rs761441714 Chr.20: 48941153-48941153 on Build GRCh38
rs768578824 Chr.20: 48941278-48941278 on Build GRCh38
rs767020632 Chr.20: 48941187-48941187 on Build GRCh38
rs769460114 Chr.20: 48941161-48941161 on Build GRCh38
rs778003839 Chr.20: 48941241-48941241 on Build GRCh38
rs752041513 Chr.20: 48941195-48941195 on Build GRCh38
rs776195775 Chr.20: 48941151-48941151 on Build GRCh38
rs759701587 Chr.20: 48941199-48941199 on Build GRCh38
rs200905854 Chr.20: 48941223-48941223 on Build GRCh38
rs779981731 Chr.20: 48941262-48941262 on Build GRCh38
rs374535118 Chr.20: 48941242-48941242 on Build GRCh38
rs1048804572 Chr.20: 48941240-48941240 on Build GRCh38
rs1054956863 Chr.20: 48941238-48941238 on Build GRCh38
rs756995732 Chr.20: 48941282-48941282 on Build GRCh38
rs1041140652 Chr.20: 48941266-48941266 on Build GRCh38
rs901295620 Chr.20: 48941293-48941293 on Build GRCh38
rs756329199 Chr.20: 48941218-48941218 on Build GRCh38
rs562273732 Chr.20: 48941251-48941251 on Build GRCh38
rs750216612 Chr.20: 48941244-48941244 on Build GRCh38
rs752815154 Chr.20: 48941217-48941217 on Build GRCh38
rs763324903 Chr.20: 48941177-48941177 on Build GRCh38
rs1000020824 Chr.20: 48941304-48941304 on Build GRCh38
rs191300623 Chr.20: 48941263-48941263 on Build GRCh38
rs865904889 Chr.20: 48941287-48941287 on Build GRCh38
rs903656897 Chr.20: 48941147-48941147 on Build GRCh38
rs200656609 Chr.20: 48941146-48941148 on Build GRCh38
rs368083068 Chr.20: 48941174-48941174 on Build GRCh38
rs767762884 Chr.20: 48941200-48941200 on Build GRCh38
rs758123963 Chr.20: 48941260-48941260 on Build GRCh38
rs1013650202 Chr.20: 48941299-48941299 on Build GRCh38

View Full Product Details