Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184261174-184261405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2N2
Entrez Gene ID
94032
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 2
Gene Aliases
CAM-KIIN, CAMKIIN
UniGene
Hs.585003
Gene Chromosome Location
Chr.3: 184259215-184261463 on Build GRCh38
Transcripts
NM_033259.2 NM_014693.3

Gene Symbol
ECE2
Entrez Gene ID
9718
Gene Name
endothelin converting enzyme 2
Gene Aliases
-
UniGene
Hs.146161
Gene Chromosome Location
Chr.3: 184249657-184293031 on Build GRCh38
Transcripts
NM_033259.2 NM_014693.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4115693 c.111C>T p.F37F Chr.3: 184261175-184261175 on Build GRCh38
1685028 c.107T>G p.F36C Chr.3: 184261179-184261179 on Build GRCh38
4876737 c.41G>T p.R14L Chr.3: 184261245-184261245 on Build GRCh38
6619230 c.109_111delTTC p.F37delF Chr.3: 184261175-184261177 on Build GRCh38
1693861 c.58G>C p.E20Q Chr.3: 184261228-184261228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766575584 Chr.3: 184261294-184261294 on Build GRCh38
rs770653095 Chr.3: 184261224-184261224 on Build GRCh38
rs554087231 Chr.3: 184261352-184261352 on Build GRCh38
rs1022281014 Chr.3: 184261264-184261264 on Build GRCh38
rs997049105 Chr.3: 184261263-184261263 on Build GRCh38
rs1008610475 Chr.3: 184261216-184261216 on Build GRCh38
rs927907434 Chr.3: 184261314-184261314 on Build GRCh38
rs200615930 Chr.3: 184261319-184261319 on Build GRCh38
rs920507614 Chr.3: 184261359-184261359 on Build GRCh38
rs973297713 Chr.3: 184261353-184261353 on Build GRCh38
rs755305063 Chr.3: 184261307-184261307 on Build GRCh38
rs772766519 Chr.3: 184261218-184261218 on Build GRCh38
rs879016909 Chr.3: 184261268-184261268 on Build GRCh38
rs567283803 Chr.3: 184261199-184261199 on Build GRCh38
rs748657032 Chr.3: 184261201-184261201 on Build GRCh38
rs781474386 Chr.3: 184261308-184261308 on Build GRCh38
rs929229713 Chr.3: 184261363-184261363 on Build GRCh38
rs753187589 Chr.3: 184261311-184261311 on Build GRCh38
rs762975012 Chr.3: 184261288-184261288 on Build GRCh38
rs770366282 Chr.3: 184261320-184261320 on Build GRCh38
rs556940963 Chr.3: 184261228-184261228 on Build GRCh38
rs914937195 Chr.3: 184261292-184261292 on Build GRCh38
rs750575240 Chr.3: 184261271-184261271 on Build GRCh38
rs751744923 Chr.3: 184261299-184261299 on Build GRCh38
rs148918121 Chr.3: 184261232-184261232 on Build GRCh38
rs755322471 Chr.3: 184261197-184261197 on Build GRCh38
rs978044883 Chr.3: 184261303-184261303 on Build GRCh38
rs539254561 Chr.3: 184261351-184261351 on Build GRCh38
rs775567043 Chr.3: 184261245-184261245 on Build GRCh38
rs765388805 Chr.3: 184261258-184261258 on Build GRCh38
rs1047352996 Chr.3: 184261381-184261381 on Build GRCh38
rs748916497 Chr.3: 184261220-184261220 on Build GRCh38
rs201603713 Chr.3: 184261217-184261217 on Build GRCh38
rs1053040629 Chr.3: 184261213-184261213 on Build GRCh38
rs756662598 Chr.3: 184261316-184261316 on Build GRCh38
rs992924012 Chr.3: 184261325-184261325 on Build GRCh38
rs777233582 Chr.3: 184261317-184261317 on Build GRCh38
rs760755093 Chr.3: 184261246-184261246 on Build GRCh38

View Full Product Details