Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27547266-27547491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHDC1
Entrez Gene ID
27245
Gene Name
AT-hook DNA binding motif containing 1
Gene Aliases
MRD25
UniGene
Hs.469280
Gene Chromosome Location
Chr.1: 27534245-27604178 on Build GRCh38
Transcripts
NM_001029882.3 XM_005245848.3 XM_005245849.3 XM_005245850.3 XM_005245851.3 XM_005245852.3 XM_011541255.1 XM_011541256.2 XM_011541257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
358089 c.4803A>T p.T1601T Chr.1: 27547313-27547313 on Build GRCh38
6540247 c.4656G>A p.P1552P Chr.1: 27547460-27547460 on Build GRCh38
4660482 c.4702G>T p.G1568C Chr.1: 27547414-27547414 on Build GRCh38
6596635 c.4782C>T p.P1594P Chr.1: 27547334-27547334 on Build GRCh38
4143760 c.4792T>G p.F1598V Chr.1: 27547324-27547324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141730488 Chr.1: 27547384-27547384 on Build GRCh38
rs570732949 Chr.1: 27547403-27547403 on Build GRCh38
rs895223934 Chr.1: 27547355-27547355 on Build GRCh38
rs769103308 Chr.1: 27547373-27547373 on Build GRCh38
rs766392554 Chr.1: 27547327-27547327 on Build GRCh38
rs61742782 Chr.1: 27547334-27547334 on Build GRCh38
rs368785856 Chr.1: 27547449-27547449 on Build GRCh38
rs150494865 Chr.1: 27547314-27547314 on Build GRCh38
rs746552201 Chr.1: 27547438-27547438 on Build GRCh38
rs766141127 Chr.1: 27547397-27547397 on Build GRCh38
rs749083604 Chr.1: 27547341-27547341 on Build GRCh38
rs779606117 Chr.1: 27547433-27547433 on Build GRCh38
rs745538283 Chr.1: 27547357-27547357 on Build GRCh38
rs771598246 Chr.1: 27547287-27547287 on Build GRCh38
rs758513170 Chr.1: 27547325-27547325 on Build GRCh38
rs773780039 Chr.1: 27547460-27547460 on Build GRCh38
rs539447084 Chr.1: 27547429-27547429 on Build GRCh38
rs763976609 Chr.1: 27547405-27547405 on Build GRCh38
rs373709770 Chr.1: 27547407-27547407 on Build GRCh38
rs776011919 Chr.1: 27547288-27547288 on Build GRCh38
rs756868783 Chr.1: 27547346-27547346 on Build GRCh38
rs866573026 Chr.1: 27547318-27547318 on Build GRCh38
rs769177974 Chr.1: 27547451-27547451 on Build GRCh38
rs774871172 Chr.1: 27547366-27547366 on Build GRCh38
rs750016023 Chr.1: 27547430-27547430 on Build GRCh38
rs371087154 Chr.1: 27547398-27547398 on Build GRCh38
rs762116470 Chr.1: 27547313-27547313 on Build GRCh38
rs760198531 Chr.1: 27547401-27547401 on Build GRCh38
rs752612737 Chr.1: 27547333-27547333 on Build GRCh38
rs770314289 Chr.1: 27547385-27547385 on Build GRCh38
rs771516746 Chr.1: 27547364-27547364 on Build GRCh38
rs756993550 Chr.1: 27547427-27547427 on Build GRCh38
rs747550510 Chr.1: 27547367-27547367 on Build GRCh38
rs745420074 Chr.1: 27547285-27547285 on Build GRCh38
rs375725827 Chr.1: 27547465-27547465 on Build GRCh38
rs138077923 Chr.1: 27547336-27547336 on Build GRCh38
rs200259294 Chr.1: 27547389-27547389 on Build GRCh38
rs774456209 Chr.1: 27547462-27547462 on Build GRCh38
rs556157635 Chr.1: 27547456-27547456 on Build GRCh38
rs149147457 Chr.1: 27547319-27547319 on Build GRCh38
rs748688517 Chr.1: 27547454-27547454 on Build GRCh38
rs141859369 Chr.1: 27547310-27547310 on Build GRCh38
rs764456894 Chr.1: 27547309-27547309 on Build GRCh38
rs529359355 Chr.1: 27547299-27547299 on Build GRCh38
rs773518872 Chr.1: 27547388-27547388 on Build GRCh38
rs778493358 Chr.1: 27547347-27547347 on Build GRCh38
rs41284274 Chr.1: 27547293-27547293 on Build GRCh38
rs781513175 Chr.1: 27547452-27547452 on Build GRCh38
rs777034610 Chr.1: 27547380-27547380 on Build GRCh38
rs753712788 Chr.1: 27547466-27547466 on Build GRCh38
rs372231427 Chr.1: 27547461-27547461 on Build GRCh38

View Full Product Details