Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63861601-63861842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD16B
Entrez Gene ID
140701
Gene Name
abhydrolase domain containing 16B
Gene Aliases
C20orf135, dJ591C20.1
UniGene
Hs.266774
Gene Chromosome Location
Chr.20: 63861213-63862988 on Build GRCh38
Transcripts
NM_080622.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5392633 c.242C>T p.A81V Chr.20: 63861623-63861623 on Build GRCh38
3707948 c.282C>T p.P94P Chr.20: 63861822-63861822 on Build GRCh38
6405527 c.88C>T p.P30S Chr.20: 63861628-63861628 on Build GRCh38
5449456 c.198G>A p.P66P Chr.20: 63861667-63861667 on Build GRCh38
6435648 c.128G>A p.R43Q Chr.20: 63861668-63861668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775224006 Chr.20: 63861745-63861745 on Build GRCh38
rs1024821218 Chr.20: 63861655-63861655 on Build GRCh38
rs965671960 Chr.20: 63861682-63861682 on Build GRCh38
rs539544290 Chr.20: 63861822-63861823 on Build GRCh38
rs765475843 Chr.20: 63861776-63861776 on Build GRCh38
rs758456348 Chr.20: 63861630-63861630 on Build GRCh38
rs770468146 Chr.20: 63861669-63861669 on Build GRCh38
rs767457694 Chr.20: 63861710-63861710 on Build GRCh38
rs949335484 Chr.20: 63861751-63861751 on Build GRCh38
rs894774514 Chr.20: 63861748-63861748 on Build GRCh38
rs999650410 Chr.20: 63861660-63861660 on Build GRCh38
rs772765062 Chr.20: 63861665-63861665 on Build GRCh38
rs557793337 Chr.20: 63861661-63861661 on Build GRCh38
rs71197437 Chr.20: 63861669-63861670 on Build GRCh38
rs540223731 Chr.20: 63861792-63861792 on Build GRCh38
rs748469189 Chr.20: 63861658-63861658 on Build GRCh38
rs755478380 Chr.20: 63861812-63861812 on Build GRCh38
rs750648123 Chr.20: 63861777-63861777 on Build GRCh38
rs766621180 Chr.20: 63861788-63861788 on Build GRCh38
rs560544546 Chr.20: 63861813-63861813 on Build GRCh38
rs112134367 Chr.20: 63861629-63861629 on Build GRCh38
rs930140698 Chr.20: 63861705-63861705 on Build GRCh38
rs941723052 Chr.20: 63861732-63861732 on Build GRCh38
rs371090962 Chr.20: 63861631-63861631 on Build GRCh38
rs370049220 Chr.20: 63861667-63861667 on Build GRCh38
rs969766501 Chr.20: 63861786-63861796 on Build GRCh38
rs747582706 Chr.20: 63861632-63861632 on Build GRCh38
rs577963504 Chr.20: 63861676-63861676 on Build GRCh38
rs908952613 Chr.20: 63861708-63861708 on Build GRCh38
rs979770892 Chr.20: 63861806-63861834 on Build GRCh38
rs781639658 Chr.20: 63861657-63861657 on Build GRCh38
rs918756050 Chr.20: 63861691-63861691 on Build GRCh38
rs201132416 Chr.20: 63861683-63861683 on Build GRCh38
rs758497660 Chr.20: 63861782-63861782 on Build GRCh38
rs755378114 Chr.20: 63861652-63861652 on Build GRCh38
rs760408112 Chr.20: 63861761-63861761 on Build GRCh38
rs748685944 Chr.20: 63861708-63861716 on Build GRCh38

View Full Product Details