Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111533645-111533892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf88
Entrez Gene ID
399949
Gene Name
chromosome 11 open reading frame 88
Gene Aliases
-
UniGene
Hs.411600
Gene Chromosome Location
Chr.11: 111514785-111537031 on Build GRCh38
Transcripts
NM_001100388.1 NM_207430.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351216 c.438G>T p.E146D Chr.11: 111533763-111533763 on Build GRCh38
5544549 c.436G>A p.E146K Chr.11: 111533761-111533761 on Build GRCh38
4017769 c.427A>G p.T143A Chr.11: 111533752-111533752 on Build GRCh38
6461514 c.458A>G p.K153R Chr.11: 111533783-111533783 on Build GRCh38
374069 c.469G>A p.E157K Chr.11: 111533794-111533794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750351101 Chr.11: 111533722-111533722 on Build GRCh38
rs545905322 Chr.11: 111533761-111533761 on Build GRCh38
rs571466596 Chr.11: 111533819-111533819 on Build GRCh38
rs112128046 Chr.11: 111533694-111533694 on Build GRCh38
rs763775143 Chr.11: 111533700-111533700 on Build GRCh38
rs760372381 Chr.11: 111533697-111533697 on Build GRCh38
rs758323194 Chr.11: 111533741-111533741 on Build GRCh38
rs201454856 Chr.11: 111533802-111533802 on Build GRCh38
rs769836561 Chr.11: 111533787-111533787 on Build GRCh38
rs1025013518 Chr.11: 111533799-111533799 on Build GRCh38
rs764879704 Chr.11: 111533721-111533721 on Build GRCh38
rs776390681 Chr.11: 111533829-111533829 on Build GRCh38
rs374370338 Chr.11: 111533775-111533775 on Build GRCh38
rs753559438 Chr.11: 111533713-111533713 on Build GRCh38
rs147855101 Chr.11: 111533694-111533697 on Build GRCh38
rs774638478 Chr.11: 111533696-111533696 on Build GRCh38
rs961237284 Chr.11: 111533691-111533691 on Build GRCh38
rs1005320014 Chr.11: 111533698-111533698 on Build GRCh38
rs1013085025 Chr.11: 111533770-111533770 on Build GRCh38
rs781338852 Chr.11: 111533772-111533772 on Build GRCh38
rs1019830720 Chr.11: 111533688-111533688 on Build GRCh38
rs61764071 Chr.11: 111533805-111533805 on Build GRCh38
rs779892648 Chr.11: 111533745-111533745 on Build GRCh38
rs370825951 Chr.11: 111533746-111533746 on Build GRCh38
rs867136385 Chr.11: 111533715-111533715 on Build GRCh38
rs766087131 Chr.11: 111533754-111533755 on Build GRCh38
rs538896697 Chr.11: 111533830-111533830 on Build GRCh38
rs933677714 Chr.11: 111533693-111533693 on Build GRCh38
rs761358403 Chr.11: 111533714-111533714 on Build GRCh38
rs200623359 Chr.11: 111533804-111533804 on Build GRCh38
rs117655744 Chr.11: 111533844-111533844 on Build GRCh38
rs902641800 Chr.11: 111533788-111533788 on Build GRCh38
rs112867149 Chr.11: 111533812-111533812 on Build GRCh38
rs762563127 Chr.11: 111533721-111533721 on Build GRCh38
rs749791327 Chr.11: 111533803-111533803 on Build GRCh38

View Full Product Details