Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86392568-86392839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC81
Entrez Gene ID
60494
Gene Name
coiled-coil domain containing 81
Gene Aliases
-
UniGene
Hs.144913
Gene Chromosome Location
Chr.11: 86374736-86423109 on Build GRCh38
Transcripts
NM_001156474.1 NM_021827.4 XM_005274161.1 XM_017018105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703726 c.473A>G p.D158G Chr.11: 86392715-86392715 on Build GRCh38
1200059 c.550G>A p.A184T Chr.11: 86392792-86392792 on Build GRCh38
5681546 c.456G>C p.G152G Chr.11: 86392698-86392698 on Build GRCh38
1579536 c.410C>T p.S137F Chr.11: 86392652-86392652 on Build GRCh38
4932556 c.426A>T p.Q142H Chr.11: 86392668-86392668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950486324 Chr.11: 86392731-86392731 on Build GRCh38
rs867090017 Chr.11: 86392679-86392679 on Build GRCh38
rs994746235 Chr.11: 86392743-86392743 on Build GRCh38
rs573544982 Chr.11: 86392727-86392727 on Build GRCh38
rs541736961 Chr.11: 86392699-86392699 on Build GRCh38
rs372945413 Chr.11: 86392646-86392646 on Build GRCh38
rs757572257 Chr.11: 86392760-86392760 on Build GRCh38
rs982295758 Chr.11: 86392604-86392604 on Build GRCh38
rs902615370 Chr.11: 86392632-86392632 on Build GRCh38
rs775327346 Chr.11: 86392629-86392629 on Build GRCh38
rs983181738 Chr.11: 86392790-86392790 on Build GRCh38
rs914954561 Chr.11: 86392643-86392643 on Build GRCh38
rs758824883 Chr.11: 86392701-86392701 on Build GRCh38
rs572643642 Chr.11: 86392695-86392695 on Build GRCh38
rs578071215 Chr.11: 86392649-86392649 on Build GRCh38
rs761458527 Chr.11: 86392647-86392647 on Build GRCh38
rs759923255 Chr.11: 86392682-86392682 on Build GRCh38
rs762713437 Chr.11: 86392631-86392631 on Build GRCh38
rs768496357 Chr.11: 86392634-86392634 on Build GRCh38
rs775314659 Chr.11: 86392637-86392637 on Build GRCh38
rs1046178056 Chr.11: 86392623-86392623 on Build GRCh38
rs749970959 Chr.11: 86392667-86392667 on Build GRCh38
rs10501615 Chr.11: 86392660-86392660 on Build GRCh38
rs558222477 Chr.11: 86392619-86392619 on Build GRCh38
rs1032326198 Chr.11: 86392636-86392637 on Build GRCh38
rs547385737 Chr.11: 86392682-86392682 on Build GRCh38
rs374558231 Chr.11: 86392728-86392728 on Build GRCh38
rs17210546 Chr.11: 86392725-86392725 on Build GRCh38

View Full Product Details