Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45650726-45650963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST1
Entrez Gene ID
8534
Gene Name
carbohydrate sulfotransferase 1
Gene Aliases
C6ST, GST-1, KS6ST, KSGal6ST, KSST
UniGene
Hs.104576
Gene Chromosome Location
Chr.11: 45647689-45665656 on Build GRCh38
Transcripts
NM_003654.5 XM_006718356.3 XM_017018459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6132809 c.40G>T p.A14S Chr.11: 45650884-45650884 on Build GRCh38
6628824 c.159C>T p.F53F Chr.11: 45650765-45650765 on Build GRCh38
3447433 c.89C>T p.S30F Chr.11: 45650835-45650835 on Build GRCh38
5372488 c.190C>T p.L64F Chr.11: 45650734-45650734 on Build GRCh38
3447434 c.69C>T p.I23I Chr.11: 45650855-45650855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747417454 Chr.11: 45650924-45650924 on Build GRCh38
rs745903357 Chr.11: 45650912-45650912 on Build GRCh38
rs769119792 Chr.11: 45650940-45650940 on Build GRCh38
rs780417954 Chr.11: 45650920-45650920 on Build GRCh38
rs567406629 Chr.11: 45650843-45650843 on Build GRCh38
rs750763320 Chr.11: 45650756-45650758 on Build GRCh38
rs140694718 Chr.11: 45650764-45650764 on Build GRCh38
rs750018576 Chr.11: 45650755-45650755 on Build GRCh38
rs776012645 Chr.11: 45650803-45650803 on Build GRCh38
rs977622491 Chr.11: 45650904-45650904 on Build GRCh38
rs746108987 Chr.11: 45650799-45650799 on Build GRCh38
rs758749450 Chr.11: 45650913-45650913 on Build GRCh38
rs145902794 Chr.11: 45650804-45650804 on Build GRCh38
rs762595086 Chr.11: 45650827-45650827 on Build GRCh38
rs757992892 Chr.11: 45650757-45650757 on Build GRCh38
rs761434923 Chr.11: 45650816-45650816 on Build GRCh38
rs550690637 Chr.11: 45650810-45650810 on Build GRCh38
rs749905203 Chr.11: 45650820-45650820 on Build GRCh38
rs764979484 Chr.11: 45650748-45650748 on Build GRCh38
rs772356003 Chr.11: 45650801-45650801 on Build GRCh38
rs769352708 Chr.11: 45650807-45650807 on Build GRCh38
rs140951795 Chr.11: 45650873-45650873 on Build GRCh38
rs779003187 Chr.11: 45650785-45650785 on Build GRCh38
rs199594578 Chr.11: 45650897-45650897 on Build GRCh38
rs766005463 Chr.11: 45650829-45650829 on Build GRCh38
rs751285636 Chr.11: 45650765-45650765 on Build GRCh38
rs968055629 Chr.11: 45650903-45650903 on Build GRCh38
rs764671809 Chr.11: 45650819-45650819 on Build GRCh38
rs878971646 Chr.11: 45650796-45650796 on Build GRCh38
rs374746953 Chr.11: 45650853-45650853 on Build GRCh38
rs200369325 Chr.11: 45650862-45650862 on Build GRCh38
rs1001961016 Chr.11: 45650790-45650790 on Build GRCh38
rs749430469 Chr.11: 45650773-45650773 on Build GRCh38
rs781106335 Chr.11: 45650772-45650772 on Build GRCh38
rs754970236 Chr.11: 45650768-45650768 on Build GRCh38
rs770897574 Chr.11: 45650779-45650779 on Build GRCh38
rs756039630 Chr.11: 45650890-45650890 on Build GRCh38

View Full Product Details