Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39229358-39229583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL4
Entrez Gene ID
344387
Gene Name
cyclin dependent kinase like 4
Gene Aliases
-
UniGene
Hs.403201
Gene Chromosome Location
Chr.2: 39174421-39246815 on Build GRCh38
Transcripts
NM_001009565.1 XM_011532815.2 XM_011532816.2 XM_011532817.2 XM_011532819.2 XM_017003981.1 XM_017003982.1 XM_017003983.1 XM_017003984.1 XM_017003985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7360684 c.158G>A p.R53H Chr.2: 39229375-39229375 on Build GRCh38
5433898 c.141A>C p.I47I Chr.2: 39229392-39229392 on Build GRCh38
7447325 c.44A>G p.Y15C Chr.2: 39229489-39229489 on Build GRCh38
4869476 c.156A>G p.I52M Chr.2: 39229377-39229377 on Build GRCh38
6488804 c.80G>A p.G27E Chr.2: 39229453-39229453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772446507 Chr.2: 39229464-39229464 on Build GRCh38
rs754737936 Chr.2: 39229400-39229400 on Build GRCh38
rs974166442 Chr.2: 39229557-39229557 on Build GRCh38
rs56089528 Chr.2: 39229458-39229458 on Build GRCh38
rs374987616 Chr.2: 39229531-39229531 on Build GRCh38
rs777011832 Chr.2: 39229488-39229488 on Build GRCh38
rs751476088 Chr.2: 39229517-39229517 on Build GRCh38
rs748763891 Chr.2: 39229552-39229552 on Build GRCh38
rs141333215 Chr.2: 39229418-39229418 on Build GRCh38
rs746455095 Chr.2: 39229445-39229445 on Build GRCh38
rs780857169 Chr.2: 39229406-39229406 on Build GRCh38
rs147155601 Chr.2: 39229492-39229492 on Build GRCh38
rs1000593915 Chr.2: 39229405-39229405 on Build GRCh38
rs758417891 Chr.2: 39229505-39229505 on Build GRCh38
rs754791362 Chr.2: 39229521-39229521 on Build GRCh38
rs755762926 Chr.2: 39229535-39229535 on Build GRCh38
rs777325599 Chr.2: 39229538-39229538 on Build GRCh38
rs199615719 Chr.2: 39229436-39229436 on Build GRCh38
rs900160261 Chr.2: 39229427-39229427 on Build GRCh38
rs35947084 Chr.2: 39229421-39229421 on Build GRCh38
rs997179174 Chr.2: 39229433-39229433 on Build GRCh38
rs770405532 Chr.2: 39229555-39229555 on Build GRCh38
rs760644652 Chr.2: 39229558-39229558 on Build GRCh38
rs772678564 Chr.2: 39229430-39229430 on Build GRCh38
rs776009745 Chr.2: 39229481-39229481 on Build GRCh38
rs149237225 Chr.2: 39229524-39229524 on Build GRCh38
rs746764745 Chr.2: 39229398-39229398 on Build GRCh38
rs765447365 Chr.2: 39229494-39229494 on Build GRCh38
rs750463961 Chr.2: 39229496-39229496 on Build GRCh38
rs138111984 Chr.2: 39229487-39229487 on Build GRCh38
rs866903206 Chr.2: 39229502-39229502 on Build GRCh38
rs766233490 Chr.2: 39229511-39229511 on Build GRCh38
rs760948791 Chr.2: 39229485-39229485 on Build GRCh38

View Full Product Details