Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99216154-99216425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH6
Entrez Gene ID
130
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
ADH-5
UniGene
Hs.586161
Gene Chromosome Location
Chr.4: 99202638-99219246 on Build GRCh38
Transcripts
NM_000672.3 NM_001102470.1 NR_132990.1 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000672.3 NM_001102470.1 NR_132990.1 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694713 c.38C>T p.A13V Chr.4: 99216243-99216243 on Build GRCh38
5558055 c.120G>A p.K40K Chr.4: 99216161-99216161 on Build GRCh38
5558056 c.120G>A p.K40K Chr.4: 99216161-99216161 on Build GRCh38
5400196 c.106G>A p.E36K Chr.4: 99216175-99216175 on Build GRCh38
6595194 c.43C>T p.L15F Chr.4: 99216238-99216238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4699733 Chr.4: 99216377-99216377 on Build GRCh38
rs779534661 Chr.4: 99216254-99216254 on Build GRCh38
rs771073132 Chr.4: 99216297-99216297 on Build GRCh38
rs755474599 Chr.4: 99216182-99216182 on Build GRCh38
rs761503688 Chr.4: 99216241-99216241 on Build GRCh38
rs202090243 Chr.4: 99216272-99216272 on Build GRCh38
rs6843298 Chr.4: 99216236-99216236 on Build GRCh38
rs140047254 Chr.4: 99216188-99216188 on Build GRCh38
rs886402796 Chr.4: 99216246-99216246 on Build GRCh38
rs940770141 Chr.4: 99216391-99216391 on Build GRCh38
rs781626054 Chr.4: 99216187-99216188 on Build GRCh38
rs769713701 Chr.4: 99216225-99216225 on Build GRCh38
rs191512358 Chr.4: 99216228-99216228 on Build GRCh38
rs753141701 Chr.4: 99216258-99216258 on Build GRCh38
rs1036175515 Chr.4: 99216394-99216394 on Build GRCh38
rs138328991 Chr.4: 99216355-99216355 on Build GRCh38
rs747254423 Chr.4: 99216281-99216281 on Build GRCh38
rs199767791 Chr.4: 99216278-99216278 on Build GRCh38
rs748489029 Chr.4: 99216189-99216189 on Build GRCh38
rs775274531 Chr.4: 99216227-99216227 on Build GRCh38
rs547103646 Chr.4: 99216353-99216353 on Build GRCh38
rs755751656 Chr.4: 99216277-99216277 on Build GRCh38
rs759396072 Chr.4: 99216212-99216212 on Build GRCh38
rs767171895 Chr.4: 99216243-99216243 on Build GRCh38
rs200929780 Chr.4: 99216233-99216233 on Build GRCh38
rs771048382 Chr.4: 99216210-99216210 on Build GRCh38
rs1019146725 Chr.4: 99216290-99216290 on Build GRCh38
rs757727115 Chr.4: 99216372-99216372 on Build GRCh38
rs749885109 Chr.4: 99216247-99216247 on Build GRCh38
rs772312242 Chr.4: 99216193-99216193 on Build GRCh38
rs936921052 Chr.4: 99216312-99216312 on Build GRCh38
rs369461150 Chr.4: 99216309-99216309 on Build GRCh38
rs199855886 Chr.4: 99216195-99216195 on Build GRCh38
rs867505885 Chr.4: 99216186-99216187 on Build GRCh38
rs778090423 Chr.4: 99216279-99216279 on Build GRCh38
rs776652444 Chr.4: 99216211-99216211 on Build GRCh38
rs749701769 Chr.4: 99216371-99216371 on Build GRCh38
rs747201312 Chr.4: 99216197-99216197 on Build GRCh38
rs990169114 Chr.4: 99216322-99216322 on Build GRCh38
rs6820368 Chr.4: 99216337-99216337 on Build GRCh38
rs80046510 Chr.4: 99216251-99216251 on Build GRCh38
rs1047740087 Chr.4: 99216183-99216183 on Build GRCh38
rs755457270 Chr.4: 99216248-99216248 on Build GRCh38
rs200690434 Chr.4: 99216380-99216381 on Build GRCh38
rs569130782 Chr.4: 99216386-99216386 on Build GRCh38
rs375353142 Chr.4: 99216300-99216300 on Build GRCh38
rs536313631 Chr.4: 99216396-99216396 on Build GRCh38
rs1051770999 Chr.4: 99216373-99216373 on Build GRCh38

View Full Product Details