Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93499919-93500174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP55
Entrez Gene ID
55165
Gene Name
centrosomal protein 55
Gene Aliases
C10orf3, CT111, URCC6
UniGene
Hs.14559
Gene Chromosome Location
Chr.10: 93496612-93529094 on Build GRCh38
Transcripts
NM_001127182.1 NM_018131.4 XM_011539918.1 XM_011539919.1 XM_011539920.2 XM_017016372.1 XM_017016373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627087 c.69C>T p.S23S Chr.10: 93500120-93500120 on Build GRCh38
6627086 c.48G>A p.S16S Chr.10: 93500099-93500099 on Build GRCh38
6627085 c.70G>A p.E24K Chr.10: 93500121-93500121 on Build GRCh38
1200826 c.85A>C p.K29Q Chr.10: 93500136-93500136 on Build GRCh38
7337372 c.8C>T p.S3F Chr.10: 93500059-93500059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931329677 Chr.10: 93499963-93499963 on Build GRCh38
rs41290212 Chr.10: 93500018-93500018 on Build GRCh38
rs746729536 Chr.10: 93500030-93500030 on Build GRCh38
rs752387704 Chr.10: 93500136-93500136 on Build GRCh38
rs558657679 Chr.10: 93500004-93500004 on Build GRCh38
rs1012047533 Chr.10: 93500056-93500056 on Build GRCh38
rs574406497 Chr.10: 93500145-93500145 on Build GRCh38
rs757917291 Chr.10: 93500023-93500023 on Build GRCh38
rs149119564 Chr.10: 93500121-93500121 on Build GRCh38
rs906393624 Chr.10: 93500094-93500094 on Build GRCh38
rs760034929 Chr.10: 93500080-93500080 on Build GRCh38
rs773616399 Chr.10: 93500047-93500047 on Build GRCh38
rs779525511 Chr.10: 93500028-93500028 on Build GRCh38
rs774728365 Chr.10: 93500078-93500078 on Build GRCh38
rs775710423 Chr.10: 93500093-93500093 on Build GRCh38
rs144885342 Chr.10: 93500069-93500069 on Build GRCh38
rs764431827 Chr.10: 93500099-93500099 on Build GRCh38
rs768265372 Chr.10: 93500046-93500046 on Build GRCh38
rs536399744 Chr.10: 93500129-93500129 on Build GRCh38
rs200933834 Chr.10: 93500127-93500127 on Build GRCh38
rs778743258 Chr.10: 93500011-93500011 on Build GRCh38
rs761515057 Chr.10: 93500114-93500114 on Build GRCh38
rs372454698 Chr.10: 93500098-93500098 on Build GRCh38
rs867508324 Chr.10: 93500061-93500061 on Build GRCh38
rs1024816244 Chr.10: 93500073-93500073 on Build GRCh38
rs749637981 Chr.10: 93500055-93500055 on Build GRCh38
rs541247110 Chr.10: 93500059-93500059 on Build GRCh38
rs754047483 Chr.10: 93500106-93500106 on Build GRCh38
rs772339514 Chr.10: 93500085-93500085 on Build GRCh38
rs765024195 Chr.10: 93500120-93500120 on Build GRCh38
rs879563505 Chr.10: 93499978-93499978 on Build GRCh38
rs960028221 Chr.10: 93500077-93500077 on Build GRCh38

View Full Product Details