Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 37266470-37266695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF4
Entrez Gene ID
56853
Gene Name
CUGBP, Elav-like family member 4
Gene Aliases
BRUNOL4, CELF-4
UniGene
Hs.435976
Gene Chromosome Location
Chr.18: 37243040-37566052 on Build GRCh38
Transcripts
NM_001025087.1 NM_001025088.1 NM_001025089.1 NM_020180.3 XM_005258308.3 XM_011526079.2 XM_011526081.2 XM_011526083.2 XM_011526084.2 XM_011526085.2 XM_011526086.2 XM_011526087.2 XM_011526090.2 XM_011526091.2 XM_011526092.2 XM_017025840.1 XM_017025841.1 XM_017025842.1 XM_017025843.1 XM_017025844.1 XM_017025845.1 XM_017025846.1 XM_017025847.1 XM_017025848.1 XM_017025849.1 XM_017025850.1 XM_017025851.1 XM_017025852.1 XM_017025853.1 XM_017025854.1 XM_017025855.1 XM_017025856.1 XM_017025857.1 XM_017025858.1 XM_017025859.1 XM_017025860.1 XM_017025861.1 XM_017025862.1 XM_017025863.1 XM_017025864.1 XM_017025865.1 XM_017025866.1 XM_017025867.1 XM_017025868.1 XM_017025869.1 XM_017025870.1 XM_017025871.1 XM_017025872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3525712 c.1124C>T p.P375L Chr.18: 37266574-37266574 on Build GRCh38
5573617 c.1165+9C>T p.? Chr.18: 37266524-37266524 on Build GRCh38
5792901 c.1165+1G>A p.? Chr.18: 37266532-37266532 on Build GRCh38
6380251 c.1129C>T p.Q377* Chr.18: 37266569-37266569 on Build GRCh38
1200685 c.1117G>A p.A373T Chr.18: 37266581-37266581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758031828 Chr.18: 37266605-37266605 on Build GRCh38
rs772877227 Chr.18: 37266537-37266537 on Build GRCh38
rs145333419 Chr.18: 37266582-37266582 on Build GRCh38
rs768110414 Chr.18: 37266520-37266520 on Build GRCh38
rs954095426 Chr.18: 37266642-37266642 on Build GRCh38
rs185520995 Chr.18: 37266518-37266518 on Build GRCh38
rs770712272 Chr.18: 37266643-37266643 on Build GRCh38
rs12458669 Chr.18: 37266536-37266536 on Build GRCh38
rs377070497 Chr.18: 37266585-37266585 on Build GRCh38
rs957422887 Chr.18: 37266528-37266528 on Build GRCh38
rs777522352 Chr.18: 37266608-37266608 on Build GRCh38
rs759568975 Chr.18: 37266557-37266557 on Build GRCh38
rs145042940 Chr.18: 37266555-37266555 on Build GRCh38
rs940399505 Chr.18: 37266669-37266669 on Build GRCh38
rs202171717 Chr.18: 37266510-37266510 on Build GRCh38
rs945600743 Chr.18: 37266493-37266493 on Build GRCh38
rs71365661 Chr.18: 37266561-37266561 on Build GRCh38
rs764301257 Chr.18: 37266499-37266499 on Build GRCh38
rs986989927 Chr.18: 37266661-37266661 on Build GRCh38
rs760568408 Chr.18: 37266543-37266543 on Build GRCh38
rs746672602 Chr.18: 37266638-37266638 on Build GRCh38
rs754319239 Chr.18: 37266584-37266584 on Build GRCh38
rs1045527012 Chr.18: 37266551-37266551 on Build GRCh38
rs371509950 Chr.18: 37266594-37266594 on Build GRCh38
rs747722508 Chr.18: 37266604-37266604 on Build GRCh38
rs747740678 Chr.18: 37266532-37266532 on Build GRCh38
rs771714885 Chr.18: 37266535-37266535 on Build GRCh38
rs767841321 Chr.18: 37266575-37266575 on Build GRCh38
rs536492294 Chr.18: 37266492-37266492 on Build GRCh38
rs1012731984 Chr.18: 37266526-37266526 on Build GRCh38
rs751784174 Chr.18: 37266506-37266506 on Build GRCh38
rs907024441 Chr.18: 37266554-37266554 on Build GRCh38
rs148935360 Chr.18: 37266567-37266567 on Build GRCh38
rs147319332 Chr.18: 37266517-37266517 on Build GRCh38
rs751698326 Chr.18: 37266558-37266558 on Build GRCh38
rs111957213 Chr.18: 37266521-37266521 on Build GRCh38
rs781639203 Chr.18: 37266512-37266512 on Build GRCh38
rs753958756 Chr.18: 37266511-37266511 on Build GRCh38
rs182148802 Chr.18: 37266581-37266581 on Build GRCh38
rs553129117 Chr.18: 37266516-37266516 on Build GRCh38
rs922164395 Chr.18: 37266580-37266580 on Build GRCh38
rs41474344 Chr.18: 37266579-37266579 on Build GRCh38
rs908977707 Chr.18: 37266597-37266597 on Build GRCh38
rs189935724 Chr.18: 37266519-37266519 on Build GRCh38

View Full Product Details