Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48634422-48634689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4426425 c.934C>A p.Q312K Chr.1: 48634510-48634510 on Build GRCh38
3355974 c.931G>A p.V311I Chr.1: 48634513-48634513 on Build GRCh38
1560577 c.869G>A p.R290H Chr.1: 48634575-48634575 on Build GRCh38
4426427 c.471C>A p.V157V Chr.1: 48634510-48634510 on Build GRCh38
1578196 c.853G>T p.G285* Chr.1: 48634591-48634591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751149612 Chr.1: 48634532-48634532 on Build GRCh38
rs1032609382 Chr.1: 48634658-48634658 on Build GRCh38
rs745710422 Chr.1: 48634514-48634514 on Build GRCh38
rs200479307 Chr.1: 48634646-48634646 on Build GRCh38
rs572399106 Chr.1: 48634465-48634465 on Build GRCh38
rs777245097 Chr.1: 48634615-48634615 on Build GRCh38
rs755023405 Chr.1: 48634563-48634563 on Build GRCh38
rs529804685 Chr.1: 48634657-48634657 on Build GRCh38
rs750094403 Chr.1: 48634634-48634634 on Build GRCh38
rs766261244 Chr.1: 48634445-48634447 on Build GRCh38
rs319983 Chr.1: 48634660-48634660 on Build GRCh38
rs763781620 Chr.1: 48634447-48634447 on Build GRCh38
rs368549103 Chr.1: 48634632-48634632 on Build GRCh38
rs756970357 Chr.1: 48634449-48634449 on Build GRCh38
rs981448845 Chr.1: 48634497-48634497 on Build GRCh38
rs368497669 Chr.1: 48634534-48634534 on Build GRCh38
rs767137782 Chr.1: 48634633-48634633 on Build GRCh38
rs200661451 Chr.1: 48634643-48634643 on Build GRCh38
rs889040279 Chr.1: 48634647-48634647 on Build GRCh38
rs747319387 Chr.1: 48634590-48634590 on Build GRCh38
rs760418748 Chr.1: 48634649-48634649 on Build GRCh38
rs866578883 Chr.1: 48634520-48634520 on Build GRCh38
rs748307842 Chr.1: 48634576-48634576 on Build GRCh38
rs370593619 Chr.1: 48634498-48634498 on Build GRCh38
rs375889126 Chr.1: 48634484-48634484 on Build GRCh38
rs771499735 Chr.1: 48634513-48634513 on Build GRCh38
rs570879614 Chr.1: 48634629-48634629 on Build GRCh38
rs768742270 Chr.1: 48634477-48634477 on Build GRCh38
rs191310576 Chr.1: 48634475-48634475 on Build GRCh38
rs1036099147 Chr.1: 48634597-48634597 on Build GRCh38
rs772408580 Chr.1: 48634580-48634580 on Build GRCh38
rs753800648 Chr.1: 48634557-48634557 on Build GRCh38
rs781087770 Chr.1: 48634456-48634456 on Build GRCh38
rs12091951 Chr.1: 48634499-48634499 on Build GRCh38
rs779969383 Chr.1: 48634470-48634470 on Build GRCh38
rs761260427 Chr.1: 48634502-48634502 on Build GRCh38
rs937571796 Chr.1: 48634536-48634536 on Build GRCh38
rs750312822 Chr.1: 48634462-48634462 on Build GRCh38
rs777863306 Chr.1: 48634581-48634581 on Build GRCh38
rs760170093 Chr.1: 48634625-48634625 on Build GRCh38
rs755914542 Chr.1: 48634553-48634553 on Build GRCh38
rs779914102 Chr.1: 48634554-48634554 on Build GRCh38
rs76634539 Chr.1: 48634479-48634479 on Build GRCh38
rs763698884 Chr.1: 48634519-48634519 on Build GRCh38
rs768036146 Chr.1: 48634626-48634626 on Build GRCh38
rs935403801 Chr.1: 48634612-48634612 on Build GRCh38
rs375888872 Chr.1: 48634602-48634602 on Build GRCh38
rs779103173 Chr.1: 48634575-48634575 on Build GRCh38

View Full Product Details