Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244552666-244552923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf101
Entrez Gene ID
257044
Gene Name
chromosome 1 open reading frame 101
Gene Aliases
-
UniGene
Hs.459534
Gene Chromosome Location
Chr.1: 244452014-244640362 on Build GRCh38
Transcripts
NM_001130957.1 NM_001242340.1 NM_173807.4 XM_011544138.2 XM_011544139.2 XM_011544142.2 XM_011544144.2 XM_011544145.2 XM_011544149.2 XM_011544151.2 XM_017000943.1 XM_017000944.1 XM_017000945.1 XM_017000946.1 XM_017000947.1 XM_017000948.1 XM_017000949.1 XM_017000950.1 XM_017000951.1 XM_017000952.1 XM_017000953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4235258 c.943A>G p.R315G Chr.1: 244552728-244552728 on Build GRCh38
1127073 c.973G>A p.G325S Chr.1: 244552758-244552758 on Build GRCh38
1340802 c.881G>A p.R294Q Chr.1: 244552666-244552666 on Build GRCh38
6062385 c.970G>T p.D324Y Chr.1: 244552755-244552755 on Build GRCh38
5750921 c.881G>T p.R294L Chr.1: 244552666-244552666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375228084 Chr.1: 244552728-244552728 on Build GRCh38
rs907958722 Chr.1: 244552809-244552809 on Build GRCh38
rs772268411 Chr.1: 244552846-244552846 on Build GRCh38
rs145403464 Chr.1: 244552786-244552786 on Build GRCh38
rs147807698 Chr.1: 244552727-244552727 on Build GRCh38
rs573139470 Chr.1: 244552717-244552717 on Build GRCh38
rs982596874 Chr.1: 244552792-244552792 on Build GRCh38
rs975310277 Chr.1: 244552750-244552750 on Build GRCh38
rs769439285 Chr.1: 244552754-244552754 on Build GRCh38
rs183851262 Chr.1: 244552843-244552843 on Build GRCh38
rs41307702 Chr.1: 244552716-244552716 on Build GRCh38
rs111806162 Chr.1: 244552751-244552751 on Build GRCh38
rs749634016 Chr.1: 244552790-244552790 on Build GRCh38
rs1032650649 Chr.1: 244552730-244552730 on Build GRCh38
rs771425574 Chr.1: 244552820-244552820 on Build GRCh38
rs775470125 Chr.1: 244552845-244552850 on Build GRCh38
rs771813567 Chr.1: 244552820-244552820 on Build GRCh38
rs920575803 Chr.1: 244552847-244552847 on Build GRCh38
rs1017608075 Chr.1: 244552823-244552823 on Build GRCh38
rs754424122 Chr.1: 244552731-244552731 on Build GRCh38
rs774685397 Chr.1: 244552825-244552825 on Build GRCh38
rs780738352 Chr.1: 244552732-244552732 on Build GRCh38

View Full Product Details