Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67520880-67521145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP2
Entrez Gene ID
51475
Gene Name
calcium binding protein 2
Gene Aliases
DFNB93
UniGene
Hs.278984
Gene Chromosome Location
Chr.11: 67518947-67523428 on Build GRCh38
Transcripts
NM_001318496.1 NM_016366.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704299 c.302G>A p.R101Q Chr.11: 67521102-67521102 on Build GRCh38
5618309 c.281G>A p.R94Q Chr.11: 67521123-67521123 on Build GRCh38
3870000 c.346G>A p.E116K Chr.11: 67521058-67521058 on Build GRCh38
3724750 c.268T>G p.F90V Chr.11: 67521136-67521136 on Build GRCh38
931125 c.364A>G p.I122V Chr.11: 67521040-67521040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371444406 Chr.11: 67521049-67521049 on Build GRCh38
rs200339707 Chr.11: 67521013-67521013 on Build GRCh38
rs376093439 Chr.11: 67521059-67521059 on Build GRCh38
rs2276118 Chr.11: 67521123-67521123 on Build GRCh38
rs754265381 Chr.11: 67521084-67521084 on Build GRCh38
rs573734434 Chr.11: 67521003-67521003 on Build GRCh38
rs776591254 Chr.11: 67520976-67520976 on Build GRCh38
rs761955590 Chr.11: 67521001-67521001 on Build GRCh38
rs201149489 Chr.11: 67521048-67521048 on Build GRCh38
rs372833735 Chr.11: 67521068-67521068 on Build GRCh38
rs868838912 Chr.11: 67521063-67521063 on Build GRCh38
rs747729576 Chr.11: 67521034-67521034 on Build GRCh38
rs950234810 Chr.11: 67520918-67520918 on Build GRCh38
rs1040390947 Chr.11: 67520981-67520981 on Build GRCh38
rs779624248 Chr.11: 67521100-67521100 on Build GRCh38
rs755561133 Chr.11: 67521025-67521025 on Build GRCh38
rs766714599 Chr.11: 67521082-67521082 on Build GRCh38
rs2276119 Chr.11: 67520977-67520977 on Build GRCh38
rs754353161 Chr.11: 67521021-67521021 on Build GRCh38
rs918884851 Chr.11: 67521117-67521117 on Build GRCh38
rs752235727 Chr.11: 67521104-67521104 on Build GRCh38
rs755540572 Chr.11: 67521085-67521085 on Build GRCh38
rs778506777 Chr.11: 67521032-67521032 on Build GRCh38
rs764113074 Chr.11: 67520978-67520978 on Build GRCh38
rs958822171 Chr.11: 67521060-67521060 on Build GRCh38
rs933021772 Chr.11: 67520941-67520941 on Build GRCh38
rs912759661 Chr.11: 67520972-67520972 on Build GRCh38
rs376870877 Chr.11: 67521047-67521047 on Build GRCh38
rs556337643 Chr.11: 67521076-67521076 on Build GRCh38
rs147530804 Chr.11: 67521014-67521014 on Build GRCh38
rs140767804 Chr.11: 67521058-67521058 on Build GRCh38
rs751653825 Chr.11: 67520982-67520982 on Build GRCh38
rs374871134 Chr.11: 67521040-67521040 on Build GRCh38
rs200631990 Chr.11: 67521122-67521122 on Build GRCh38
rs201625554 Chr.11: 67521105-67521105 on Build GRCh38
rs139895134 Chr.11: 67521124-67521124 on Build GRCh38
rs963261198 Chr.11: 67521102-67521102 on Build GRCh38
rs771613171 Chr.11: 67521043-67521043 on Build GRCh38
rs368368611 Chr.11: 67521065-67521065 on Build GRCh38
rs79762021 Chr.11: 67520955-67520955 on Build GRCh38
rs760834323 Chr.11: 67521081-67521081 on Build GRCh38
rs771436290 Chr.11: 67520928-67520928 on Build GRCh38
rs747312805 Chr.11: 67520915-67520915 on Build GRCh38
rs769729934 Chr.11: 67521064-67521064 on Build GRCh38
rs746624712 Chr.11: 67521118-67521118 on Build GRCh38
rs144698936 Chr.11: 67521119-67521119 on Build GRCh38
rs780390227 Chr.11: 67521017-67521017 on Build GRCh38
rs750694651 Chr.11: 67521077-67521077 on Build GRCh38
rs778281943 Chr.11: 67520908-67520908 on Build GRCh38

View Full Product Details