Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1012464-1012729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233123 c.662G>A p.R221H Chr.19: 1012519-1012519 on Build GRCh38
7394285 c.649T>A p.Y217N Chr.19: 1012532-1012532 on Build GRCh38
4424137 c.676A>C p.T226P Chr.19: 1012505-1012505 on Build GRCh38
5516797 c.700G>A p.V234I Chr.19: 1012481-1012481 on Build GRCh38
4778574 c.608-9G>T p.? Chr.19: 1012582-1012582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771560080 Chr.19: 1012567-1012567 on Build GRCh38
rs774112698 Chr.19: 1012495-1012495 on Build GRCh38
rs757361940 Chr.19: 1012551-1012551 on Build GRCh38
rs1025165774 Chr.19: 1012581-1012581 on Build GRCh38
rs754357918 Chr.19: 1012618-1012618 on Build GRCh38
rs772542168 Chr.19: 1012512-1012512 on Build GRCh38
rs1042312675 Chr.19: 1012686-1012686 on Build GRCh38
rs1009507476 Chr.19: 1012506-1012506 on Build GRCh38
rs141106687 Chr.19: 1012584-1012584 on Build GRCh38
rs772849765 Chr.19: 1012568-1012568 on Build GRCh38
rs922002452 Chr.19: 1012630-1012630 on Build GRCh38
rs781334420 Chr.19: 1012554-1012554 on Build GRCh38
rs1019038654 Chr.19: 1012641-1012641 on Build GRCh38
rs758740479 Chr.19: 1012532-1012532 on Build GRCh38
rs751852417 Chr.19: 1012594-1012594 on Build GRCh38
rs189242035 Chr.19: 1012679-1012679 on Build GRCh38
rs764466375 Chr.19: 1012590-1012590 on Build GRCh38
rs780113649 Chr.19: 1012562-1012562 on Build GRCh38
rs764341636 Chr.19: 1012539-1012539 on Build GRCh38
rs957833025 Chr.19: 1012704-1012704 on Build GRCh38
rs116349199 Chr.19: 1012566-1012566 on Build GRCh38
rs61743931 Chr.19: 1012560-1012560 on Build GRCh38
rs763411583 Chr.19: 1012589-1012589 on Build GRCh38
rs116496381 Chr.19: 1012542-1012542 on Build GRCh38
rs1050975566 Chr.19: 1012482-1012482 on Build GRCh38
rs970100 Chr.19: 1012612-1012612 on Build GRCh38
rs950705142 Chr.19: 1012626-1012626 on Build GRCh38
rs1051350974 Chr.19: 1012654-1012654 on Build GRCh38
rs750564631 Chr.19: 1012604-1012604 on Build GRCh38
rs765662869 Chr.19: 1012520-1012520 on Build GRCh38
rs761515645 Chr.19: 1012502-1012502 on Build GRCh38
rs767620474 Chr.19: 1012603-1012603 on Build GRCh38
rs369871414 Chr.19: 1012615-1012615 on Build GRCh38
rs749131136 Chr.19: 1012565-1012565 on Build GRCh38
rs71335287 Chr.19: 1012597-1012597 on Build GRCh38
rs903539756 Chr.19: 1012519-1012519 on Build GRCh38
rs539800238 Chr.19: 1012702-1012702 on Build GRCh38
rs990375019 Chr.19: 1012706-1012706 on Build GRCh38
rs373728375 Chr.19: 1012616-1012616 on Build GRCh38
rs62131163 Chr.19: 1012640-1012640 on Build GRCh38
rs745931155 Chr.19: 1012559-1012559 on Build GRCh38
rs756112380 Chr.19: 1012607-1012607 on Build GRCh38
rs767074289 Chr.19: 1012505-1012505 on Build GRCh38
rs760093321 Chr.19: 1012580-1012580 on Build GRCh38
rs974789440 Chr.19: 1012623-1012623 on Build GRCh38
rs775630468 Chr.19: 1012649-1012649 on Build GRCh38
rs966189158 Chr.19: 1012655-1012655 on Build GRCh38
rs760096106 Chr.19: 1012518-1012518 on Build GRCh38
rs553341186 Chr.19: 1012550-1012550 on Build GRCh38
rs770383050 Chr.19: 1012583-1012583 on Build GRCh38
rs753122096 Chr.19: 1012524-1012524 on Build GRCh38
rs995458002 Chr.19: 1012609-1012609 on Build GRCh38

View Full Product Details