Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57232477-57232732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKC
Entrez Gene ID
6795
Gene Name
aurora kinase C
Gene Aliases
AIE2, AIK3, ARK3, AurC, HEL-S-90, SPGF5, STK13, aurora-C
UniGene
Hs.98338
Gene Chromosome Location
Chr.19: 57230317-57235550 on Build GRCh38
Transcripts
NM_001015878.1 NM_001015879.1 NM_003160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3540371 c.207C>T p.I69I Chr.19: 57232554-57232554 on Build GRCh38
6243059 c.426C>T p.R142R Chr.19: 57232671-57232671 on Build GRCh38
4991351 c.259G>A p.E87K Chr.19: 57232606-57232606 on Build GRCh38
6152655 c.423G>C p.Q141H Chr.19: 57232668-57232668 on Build GRCh38
2971878 c.405C>T p.S135S Chr.19: 57232650-57232650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762160840 Chr.19: 57232615-57232615 on Build GRCh38
rs564437807 Chr.19: 57232634-57232634 on Build GRCh38
rs1039212821 Chr.19: 57232565-57232565 on Build GRCh38
rs777649755 Chr.19: 57232576-57232576 on Build GRCh38
rs45623632 Chr.19: 57232571-57232571 on Build GRCh38
rs549736371 Chr.19: 57232651-57232651 on Build GRCh38
rs754049495 Chr.19: 57232563-57232563 on Build GRCh38
rs202030166 Chr.19: 57232549-57232549 on Build GRCh38
rs770575687 Chr.19: 57232504-57232504 on Build GRCh38
rs929295265 Chr.19: 57232531-57232531 on Build GRCh38
rs547639001 Chr.19: 57232589-57232589 on Build GRCh38
rs776211687 Chr.19: 57232510-57232510 on Build GRCh38
rs750898508 Chr.19: 57232626-57232626 on Build GRCh38
rs763271195 Chr.19: 57232545-57232545 on Build GRCh38
rs201284711 Chr.19: 57232502-57232502 on Build GRCh38
rs527988086 Chr.19: 57232527-57232527 on Build GRCh38
rs199855150 Chr.19: 57232577-57232577 on Build GRCh38
rs372025800 Chr.19: 57232707-57232707 on Build GRCh38
rs750569894 Chr.19: 57232555-57232555 on Build GRCh38
rs142933103 Chr.19: 57232548-57232548 on Build GRCh38
rs370357811 Chr.19: 57232550-57232550 on Build GRCh38
rs748898565 Chr.19: 57232690-57232690 on Build GRCh38
rs193015358 Chr.19: 57232691-57232691 on Build GRCh38
rs768917128 Chr.19: 57232610-57232610 on Build GRCh38
rs781321656 Chr.19: 57232670-57232670 on Build GRCh38
rs772384654 Chr.19: 57232710-57232710 on Build GRCh38
rs45503793 Chr.19: 57232679-57232679 on Build GRCh38
rs368597257 Chr.19: 57232689-57232689 on Build GRCh38
rs762620183 Chr.19: 57232603-57232603 on Build GRCh38
rs759870933 Chr.19: 57232519-57232520 on Build GRCh38
rs775038920 Chr.19: 57232599-57232599 on Build GRCh38
rs765979241 Chr.19: 57232505-57232508 on Build GRCh38
rs774633250 Chr.19: 57232613-57232613 on Build GRCh38
rs769321124 Chr.19: 57232591-57232591 on Build GRCh38
rs779269368 Chr.19: 57232658-57232658 on Build GRCh38
rs374092036 Chr.19: 57232500-57232500 on Build GRCh38
rs767588611 Chr.19: 57232553-57232553 on Build GRCh38
rs757093824 Chr.19: 57232669-57232669 on Build GRCh38
rs757432491 Chr.19: 57232708-57232708 on Build GRCh38
rs769213549 Chr.19: 57232680-57232680 on Build GRCh38
rs759108151 Chr.19: 57232518-57232518 on Build GRCh38
rs771054259 Chr.19: 57232582-57232582 on Build GRCh38
rs919273503 Chr.19: 57232506-57232506 on Build GRCh38
rs1051922045 Chr.19: 57232586-57232586 on Build GRCh38
rs376035876 Chr.19: 57232514-57232514 on Build GRCh38
rs781258727 Chr.19: 57232588-57232588 on Build GRCh38
rs886054646 Chr.19: 57232596-57232596 on Build GRCh38
rs968393333 Chr.19: 57232525-57232525 on Build GRCh38
rs754153447 Chr.19: 57232511-57232516 on Build GRCh38
rs1049635989 Chr.19: 57232538-57232538 on Build GRCh38
rs141028635 Chr.19: 57232650-57232650 on Build GRCh38
rs1053349926 Chr.19: 57232585-57232585 on Build GRCh38
rs756195705 Chr.19: 57232558-57232558 on Build GRCh38
rs764851783 Chr.19: 57232519-57232519 on Build GRCh38
rs758149112 Chr.19: 57232567-57232567 on Build GRCh38
rs979438570 Chr.19: 57232528-57232528 on Build GRCh38
rs768071401 Chr.19: 57232620-57232620 on Build GRCh38
rs199610640 Chr.19: 57232501-57232501 on Build GRCh38
rs748043145 Chr.19: 57232640-57232640 on Build GRCh38
rs55710619 Chr.19: 57232641-57232641 on Build GRCh38
rs45555141 Chr.19: 57232559-57232559 on Build GRCh38

View Full Product Details