Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78113899-78114124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
UniGene
Hs.503368
Gene Chromosome Location
Chr.11: 78100942-78139660 on Build GRCh38
Transcripts
NM_001007027.2 NM_024079.4 XM_005274247.3 XM_011545252.2 XM_017018274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
932242 c.738C>A p.F246L Chr.11: 78113925-78113925 on Build GRCh38
196637 c.709C>T p.R237C Chr.11: 78113954-78113954 on Build GRCh38
6327405 c.686G>T p.R229L Chr.11: 78113977-78113977 on Build GRCh38
6317078 c.685C>A p.R229R Chr.11: 78113978-78113978 on Build GRCh38
1605262 c.722T>C p.L241P Chr.11: 78113941-78113941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754476593 Chr.11: 78113985-78113985 on Build GRCh38
rs141280441 Chr.11: 78113962-78113962 on Build GRCh38
rs533514438 Chr.11: 78114011-78114011 on Build GRCh38
rs756061099 Chr.11: 78113977-78113977 on Build GRCh38
rs1056292840 Chr.11: 78114084-78114084 on Build GRCh38
rs533704173 Chr.11: 78113978-78113978 on Build GRCh38
rs764263526 Chr.11: 78113956-78113956 on Build GRCh38
rs760030470 Chr.11: 78113934-78113934 on Build GRCh38
rs750393405 Chr.11: 78113963-78113963 on Build GRCh38
rs759036484 Chr.11: 78114072-78114072 on Build GRCh38
rs775945856 Chr.11: 78114031-78114031 on Build GRCh38
rs199603474 Chr.11: 78113996-78113996 on Build GRCh38
rs775490122 Chr.11: 78113951-78113951 on Build GRCh38
rs1003406006 Chr.11: 78114080-78114080 on Build GRCh38
rs924711179 Chr.11: 78114047-78114047 on Build GRCh38
rs572693 Chr.11: 78114018-78114018 on Build GRCh38
rs763258107 Chr.11: 78113953-78113953 on Build GRCh38
rs61566223 Chr.11: 78114020-78114020 on Build GRCh38
rs759045647 Chr.11: 78114027-78114034 on Build GRCh38
rs11237397 Chr.11: 78114016-78114016 on Build GRCh38
rs769865988 Chr.11: 78113944-78113944 on Build GRCh38
rs547807006 Chr.11: 78113968-78113968 on Build GRCh38
rs762991744 Chr.11: 78114036-78114036 on Build GRCh38
rs139106381 Chr.11: 78113988-78113988 on Build GRCh38
rs751783361 Chr.11: 78113961-78113961 on Build GRCh38
rs150736564 Chr.11: 78113981-78113981 on Build GRCh38
rs978844670 Chr.11: 78114035-78114035 on Build GRCh38
rs750895613 Chr.11: 78113938-78113938 on Build GRCh38
rs555118480 Chr.11: 78113932-78113932 on Build GRCh38
rs768929600 Chr.11: 78114050-78114050 on Build GRCh38
rs371571471 Chr.11: 78114009-78114009 on Build GRCh38
rs370235246 Chr.11: 78113928-78113928 on Build GRCh38
rs777178184 Chr.11: 78113931-78113931 on Build GRCh38
rs777227210 Chr.11: 78114088-78114088 on Build GRCh38
rs780673624 Chr.11: 78114083-78114083 on Build GRCh38
rs886287555 Chr.11: 78114073-78114073 on Build GRCh38

View Full Product Details