Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174753324-174753581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA1
Entrez Gene ID
1134
Gene Name
cholinergic receptor nicotinic alpha 1 subunit
Gene Aliases
ACHRA, ACHRD, CHRNA, CMS1A, CMS1B, CMS2A, FCCMS, SCCMS
UniGene
Hs.434479
Gene Chromosome Location
Chr.2: 174747592-174764472 on Build GRCh38
Transcripts
NM_000079.3 NM_001039523.2 XM_017003256.1 XM_017003257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4508950 c.792C>T p.I264I Chr.2: 174753564-174753564 on Build GRCh38
7094126 c.777C>A p.F259L Chr.2: 174753579-174753579 on Build GRCh38
717687 c.795C>A p.I265I Chr.2: 174753561-174753561 on Build GRCh38
1129495 c.786C>T p.N262N Chr.2: 174753570-174753570 on Build GRCh38
2894130 c.791T>G p.I264S Chr.2: 174753565-174753565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753193 Chr.2: 174753358-174753358 on Build GRCh38
rs751447629 Chr.2: 174753469-174753469 on Build GRCh38
rs750740573 Chr.2: 174753396-174753396 on Build GRCh38
rs189442556 Chr.2: 174753558-174753558 on Build GRCh38
rs186607397 Chr.2: 174753531-174753531 on Build GRCh38
rs370000238 Chr.2: 174753421-174753421 on Build GRCh38
rs752126174 Chr.2: 174753449-174753449 on Build GRCh38
rs765432113 Chr.2: 174753427-174753427 on Build GRCh38
rs554506973 Chr.2: 174753359-174753359 on Build GRCh38
rs755402316 Chr.2: 174753453-174753453 on Build GRCh38
rs766474459 Chr.2: 174753552-174753552 on Build GRCh38
rs1048719302 Chr.2: 174753376-174753376 on Build GRCh38
rs1017622034 Chr.2: 174753495-174753495 on Build GRCh38
rs374516609 Chr.2: 174753479-174753479 on Build GRCh38
rs891668768 Chr.2: 174753445-174753445 on Build GRCh38
rs938178855 Chr.2: 174753394-174753394 on Build GRCh38
rs546162718 Chr.2: 174753397-174753397 on Build GRCh38
rs765523842 Chr.2: 174753368-174753368 on Build GRCh38
rs373695811 Chr.2: 174753354-174753354 on Build GRCh38
rs137852805 Chr.2: 174753524-174753524 on Build GRCh38
rs201897865 Chr.2: 174753454-174753454 on Build GRCh38
rs926825470 Chr.2: 174753392-174753392 on Build GRCh38
rs201033430 Chr.2: 174753480-174753480 on Build GRCh38
rs752951145 Chr.2: 174753456-174753456 on Build GRCh38
rs745821257 Chr.2: 174753499-174753499 on Build GRCh38
rs180691848 Chr.2: 174753402-174753402 on Build GRCh38
rs375471139 Chr.2: 174753485-174753485 on Build GRCh38
rs483353046 Chr.2: 174753544-174753544 on Build GRCh38
rs367616114 Chr.2: 174753470-174753470 on Build GRCh38
rs903853714 Chr.2: 174753430-174753430 on Build GRCh38
rs117100375 Chr.2: 174753406-174753406 on Build GRCh38
rs372014043 Chr.2: 174753516-174753516 on Build GRCh38
rs773397465 Chr.2: 174753546-174753546 on Build GRCh38
rs76415154 Chr.2: 174753375-174753375 on Build GRCh38
rs533985924 Chr.2: 174753401-174753401 on Build GRCh38
rs770040016 Chr.2: 174753535-174753535 on Build GRCh38
rs771845966 Chr.2: 174753505-174753505 on Build GRCh38
rs750568031 Chr.2: 174753437-174753437 on Build GRCh38
rs575920001 Chr.2: 174753357-174753357 on Build GRCh38
rs367631396 Chr.2: 174753457-174753457 on Build GRCh38
rs777563775 Chr.2: 174753459-174753459 on Build GRCh38
rs544771332 Chr.2: 174753419-174753419 on Build GRCh38
rs780008784 Chr.2: 174753443-174753443 on Build GRCh38

View Full Product Details