Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186385977-186386232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
UniGene
Hs.371210
Gene Chromosome Location
Chr.1: 186375758-186421375 on Build GRCh38
Transcripts
NM_001164245.1 NM_001164246.1 NM_017847.5 XM_011509660.1 XM_011509661.1 XM_017001543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349900 c.300G>T p.L100L Chr.1: 186386053-186386053 on Build GRCh38
4633450 c.253G>T p.G85W Chr.1: 186386006-186386006 on Build GRCh38
86884 c.279T>C p.F93F Chr.1: 186386032-186386032 on Build GRCh38
4414361 c.309T>C p.D103D Chr.1: 186386062-186386062 on Build GRCh38
4789856 c.275T>C p.V92A Chr.1: 186386028-186386028 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1048897301 Chr.1: 186386034-186386034 on Build GRCh38
rs750735676 Chr.1: 186386078-186386078 on Build GRCh38
rs752881893 Chr.1: 186386099-186386099 on Build GRCh38
rs942959169 Chr.1: 186386037-186386037 on Build GRCh38
rs971919662 Chr.1: 186386187-186386187 on Build GRCh38
rs754371454 Chr.1: 186386086-186386086 on Build GRCh38
rs779007486 Chr.1: 186386094-186386094 on Build GRCh38
rs775727262 Chr.1: 186386128-186386140 on Build GRCh38
rs768075056 Chr.1: 186386130-186386130 on Build GRCh38
rs115570554 Chr.1: 186386036-186386036 on Build GRCh38
rs762499973 Chr.1: 186386033-186386033 on Build GRCh38
rs528881905 Chr.1: 186386047-186386047 on Build GRCh38
rs769562970 Chr.1: 186386124-186386124 on Build GRCh38
rs373325018 Chr.1: 186386119-186386119 on Build GRCh38
rs761790195 Chr.1: 186386125-186386125 on Build GRCh38
rs769652824 Chr.1: 186386016-186386016 on Build GRCh38
rs1041745997 Chr.1: 186386107-186386107 on Build GRCh38
rs767055367 Chr.1: 186386080-186386080 on Build GRCh38
rs761479865 Chr.1: 186386043-186386043 on Build GRCh38
rs780989697 Chr.1: 186386009-186386009 on Build GRCh38
rs897621819 Chr.1: 186386185-186386185 on Build GRCh38
rs755144990 Chr.1: 186386093-186386093 on Build GRCh38
rs551433981 Chr.1: 186386184-186386184 on Build GRCh38
rs758586395 Chr.1: 186386100-186386100 on Build GRCh38
rs961954033 Chr.1: 186386163-186386163 on Build GRCh38
rs139713643 Chr.1: 186386079-186386079 on Build GRCh38
rs773909314 Chr.1: 186386042-186386042 on Build GRCh38
rs931400747 Chr.1: 186386025-186386025 on Build GRCh38
rs771831128 Chr.1: 186386122-186386123 on Build GRCh38
rs745816508 Chr.1: 186386015-186386015 on Build GRCh38
rs539132507 Chr.1: 186386129-186386129 on Build GRCh38
rs775401978 Chr.1: 186386018-186386018 on Build GRCh38
rs565782442 Chr.1: 186386123-186386123 on Build GRCh38

View Full Product Details