Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113428841-113429068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL1
Entrez Gene ID
113622
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
ARH2
UniGene
Hs.98669
Gene Chromosome Location
Chr.13: 113421940-113453524 on Build GRCh38
Transcripts
NM_138430.4 NM_199162.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4166698 c.634C>T p.R212W Chr.13: 113428964-113428964 on Build GRCh38
5550883 c.620G>C p.C207S Chr.13: 113428978-113428978 on Build GRCh38
4503046 c.629C>T p.T210I Chr.13: 113428969-113428969 on Build GRCh38
4046199 c.530C>G p.A177G Chr.13: 113429068-113429068 on Build GRCh38
1244526 c.642G>A p.T214T Chr.13: 113428956-113428956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050676645 Chr.13: 113429017-113429017 on Build GRCh38
rs181386121 Chr.13: 113429002-113429002 on Build GRCh38
rs890655470 Chr.13: 113428885-113428885 on Build GRCh38
rs907962027 Chr.13: 113428989-113428989 on Build GRCh38
rs769603778 Chr.13: 113429037-113429041 on Build GRCh38
rs1008620939 Chr.13: 113428889-113428889 on Build GRCh38
rs763157942 Chr.13: 113429000-113429000 on Build GRCh38
rs920619446 Chr.13: 113429013-113429013 on Build GRCh38
rs372696352 Chr.13: 113428945-113428945 on Build GRCh38
rs750172361 Chr.13: 113429020-113429020 on Build GRCh38
rs139697294 Chr.13: 113429015-113429015 on Build GRCh38
rs142661838 Chr.13: 113428978-113428978 on Build GRCh38
rs377658823 Chr.13: 113429048-113429048 on Build GRCh38
rs776760378 Chr.13: 113428920-113428920 on Build GRCh38
rs190623578 Chr.13: 113428976-113428976 on Build GRCh38
rs928608999 Chr.13: 113428872-113428872 on Build GRCh38
rs760073697 Chr.13: 113428937-113428937 on Build GRCh38
rs563174718 Chr.13: 113428957-113428957 on Build GRCh38
rs947577966 Chr.13: 113428933-113428933 on Build GRCh38
rs748290770 Chr.13: 113428979-113428979 on Build GRCh38
rs111764520 Chr.13: 113428935-113428935 on Build GRCh38
rs199692255 Chr.13: 113429005-113429005 on Build GRCh38
rs200118384 Chr.13: 113428941-113428941 on Build GRCh38
rs769817799 Chr.13: 113428983-113428983 on Build GRCh38
rs932206858 Chr.13: 113429016-113429016 on Build GRCh38
rs754691717 Chr.13: 113429023-113429023 on Build GRCh38
rs746937234 Chr.13: 113428912-113428912 on Build GRCh38
rs199557593 Chr.13: 113428964-113428964 on Build GRCh38
rs773462863 Chr.13: 113428994-113428994 on Build GRCh38
rs758114074 Chr.13: 113429022-113429022 on Build GRCh38
rs775546415 Chr.13: 113428934-113428935 on Build GRCh38
rs756251846 Chr.13: 113429035-113429035 on Build GRCh38
rs764812133 Chr.13: 113429018-113429018 on Build GRCh38
rs372913606 Chr.13: 113428926-113428926 on Build GRCh38
rs9635114 Chr.13: 113429001-113429001 on Build GRCh38
rs375108772 Chr.13: 113429006-113429006 on Build GRCh38
rs1009069798 Chr.13: 113429044-113429044 on Build GRCh38
rs377490207 Chr.13: 113428966-113428966 on Build GRCh38
rs780193007 Chr.13: 113428910-113428910 on Build GRCh38
rs201361406 Chr.13: 113428936-113428936 on Build GRCh38
rs748045770 Chr.13: 113429029-113429029 on Build GRCh38
rs778727540 Chr.13: 113428955-113428955 on Build GRCh38
rs774995395 Chr.13: 113428956-113428956 on Build GRCh38
rs753094234 Chr.13: 113429011-113429011 on Build GRCh38
rs749307877 Chr.13: 113429049-113429049 on Build GRCh38
rs371692782 Chr.13: 113428925-113428925 on Build GRCh38
rs760849716 Chr.13: 113428992-113428993 on Build GRCh38

View Full Product Details