Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56933290-56933563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
UniGene
Hs.391835
Gene Chromosome Location
Chr.1: 56929210-56974383 on Build GRCh38
Transcripts
NM_000066.3 NM_001278543.1 NM_001278544.1 XM_017002235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3865883 c.1534G>A p.G512R Chr.1: 56933353-56933353 on Build GRCh38
5517930 c.1445C>G p.S482C Chr.1: 56933442-56933442 on Build GRCh38
3865884 c.1445C>T p.S482F Chr.1: 56933442-56933442 on Build GRCh38
5847060 c.1552G>A p.G518R Chr.1: 56933335-56933335 on Build GRCh38
911103 c.1552+2T>C p.? Chr.1: 56933333-56933333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959762938 Chr.1: 56933322-56933322 on Build GRCh38
rs770783602 Chr.1: 56933463-56933463 on Build GRCh38
rs757472806 Chr.1: 56933508-56933508 on Build GRCh38
rs767062675 Chr.1: 56933337-56933337 on Build GRCh38
rs374016607 Chr.1: 56933447-56933447 on Build GRCh38
rs565926603 Chr.1: 56933485-56933485 on Build GRCh38
rs747130071 Chr.1: 56933409-56933409 on Build GRCh38
rs140570606 Chr.1: 56933429-56933429 on Build GRCh38
rs749145544 Chr.1: 56933373-56933373 on Build GRCh38
rs754941165 Chr.1: 56933322-56933323 on Build GRCh38
rs374104718 Chr.1: 56933526-56933526 on Build GRCh38
rs759525365 Chr.1: 56933484-56933484 on Build GRCh38
rs745932084 Chr.1: 56933524-56933524 on Build GRCh38
rs751695720 Chr.1: 56933506-56933506 on Build GRCh38
rs1017237351 Chr.1: 56933325-56933325 on Build GRCh38
rs554618167 Chr.1: 56933450-56933450 on Build GRCh38
rs746419831 Chr.1: 56933431-56933431 on Build GRCh38
rs754881228 Chr.1: 56933383-56933383 on Build GRCh38
rs760046072 Chr.1: 56933351-56933351 on Build GRCh38
rs756354353 Chr.1: 56933464-56933464 on Build GRCh38
rs1049800151 Chr.1: 56933492-56933492 on Build GRCh38
rs781472968 Chr.1: 56933512-56933512 on Build GRCh38
rs1040882822 Chr.1: 56933470-56933470 on Build GRCh38
rs778732160 Chr.1: 56933390-56933390 on Build GRCh38
rs753735515 Chr.1: 56933371-56933371 on Build GRCh38
rs765845132 Chr.1: 56933479-56933479 on Build GRCh38
rs142870813 Chr.1: 56933482-56933482 on Build GRCh38
rs770404146 Chr.1: 56933436-56933436 on Build GRCh38
rs1005527769 Chr.1: 56933531-56933531 on Build GRCh38
rs774396158 Chr.1: 56933505-56933505 on Build GRCh38
rs924523629 Chr.1: 56933449-56933449 on Build GRCh38
rs758067323 Chr.1: 56933502-56933502 on Build GRCh38
rs766363026 Chr.1: 56933358-56933358 on Build GRCh38
rs770871056 Chr.1: 56933420-56933420 on Build GRCh38
rs373866445 Chr.1: 56933445-56933445 on Build GRCh38
rs777795608 Chr.1: 56933399-56933399 on Build GRCh38
rs760262006 Chr.1: 56933473-56933473 on Build GRCh38
rs752490325 Chr.1: 56933488-56933488 on Build GRCh38
rs764403758 Chr.1: 56933504-56933504 on Build GRCh38
rs767536852 Chr.1: 56933346-56933346 on Build GRCh38
rs200245622 Chr.1: 56933395-56933395 on Build GRCh38
rs776057562 Chr.1: 56933442-56933442 on Build GRCh38
rs761605590 Chr.1: 56933331-56933331 on Build GRCh38
rs867309304 Chr.1: 56933341-56933341 on Build GRCh38
rs752754320 Chr.1: 56933446-56933446 on Build GRCh38
rs750015764 Chr.1: 56933347-56933347 on Build GRCh38
rs749685215 Chr.1: 56933528-56933528 on Build GRCh38
rs749667478 Chr.1: 56933444-56933444 on Build GRCh38

View Full Product Details