Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93242749-93242982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
UniGene
Hs.525549
Gene Chromosome Location
Chr.14: 93237550-93333092 on Build GRCh38
Transcripts
NM_001002860.3 NM_001289133.1 XM_011536939.2 XM_017021438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7412305 c.1738C>G p.Q580E Chr.14: 93242881-93242881 on Build GRCh38
470358 c.2827C>G p.P943A Chr.14: 93242845-93242845 on Build GRCh38
1371644 c.2778_2779insA p.H927fs*21 Chr.14: 93242893-93242894 on Build GRCh38
6613412 c.2771G>A p.R924Q Chr.14: 93242901-93242901 on Build GRCh38
6613396 c.1705A>T p.T569S Chr.14: 93242914-93242914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773536280 Chr.14: 93242949-93242949 on Build GRCh38
rs745694971 Chr.14: 93242901-93242901 on Build GRCh38
rs769633347 Chr.14: 93242792-93242792 on Build GRCh38
rs762097299 Chr.14: 93242821-93242821 on Build GRCh38
rs755055327 Chr.14: 93242910-93242910 on Build GRCh38
rs1017294619 Chr.14: 93242882-93242882 on Build GRCh38
rs755765694 Chr.14: 93242902-93242902 on Build GRCh38
rs761078695 Chr.14: 93242955-93242955 on Build GRCh38
rs751550851 Chr.14: 93242872-93242872 on Build GRCh38
rs569825061 Chr.14: 93242934-93242934 on Build GRCh38
rs1014197908 Chr.14: 93242785-93242785 on Build GRCh38
rs753934329 Chr.14: 93242778-93242778 on Build GRCh38
rs775030206 Chr.14: 93242794-93242794 on Build GRCh38
rs758327406 Chr.14: 93242782-93242782 on Build GRCh38
rs748789467 Chr.14: 93242911-93242911 on Build GRCh38
rs753933887 Chr.14: 93242855-93242855 on Build GRCh38
rs200000140 Chr.14: 93242824-93242824 on Build GRCh38
rs1055837162 Chr.14: 93242781-93242781 on Build GRCh38
rs537138286 Chr.14: 93242773-93242773 on Build GRCh38
rs760882700 Chr.14: 93242828-93242828 on Build GRCh38
rs539024790 Chr.14: 93242890-93242890 on Build GRCh38
rs773624356 Chr.14: 93242826-93242826 on Build GRCh38
rs375048606 Chr.14: 93242800-93242800 on Build GRCh38
rs951864072 Chr.14: 93242848-93242848 on Build GRCh38
rs993295405 Chr.14: 93242832-93242832 on Build GRCh38
rs745533607 Chr.14: 93242790-93242790 on Build GRCh38
rs772633702 Chr.14: 93242936-93242936 on Build GRCh38
rs139558603 Chr.14: 93242774-93242774 on Build GRCh38
rs769155556 Chr.14: 93242807-93242807 on Build GRCh38
rs774750400 Chr.14: 93242816-93242816 on Build GRCh38
rs774862562 Chr.14: 93242933-93242933 on Build GRCh38
rs755038079 Chr.14: 93242861-93242861 on Build GRCh38
rs868552884 Chr.14: 93242956-93242956 on Build GRCh38
rs201926967 Chr.14: 93242784-93242784 on Build GRCh38
rs764055509 Chr.14: 93242871-93242871 on Build GRCh38
rs768054452 Chr.14: 93242921-93242921 on Build GRCh38
rs779592469 Chr.14: 93242909-93242909 on Build GRCh38
rs570431979 Chr.14: 93242835-93242835 on Build GRCh38
rs780929084 Chr.14: 93242787-93242787 on Build GRCh38
rs367623164 Chr.14: 93242916-93242916 on Build GRCh38
rs960276141 Chr.14: 93242795-93242795 on Build GRCh38
rs757065285 Chr.14: 93242875-93242875 on Build GRCh38

View Full Product Details