Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103216119-103216390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEX4
Entrez Gene ID
56271
Gene Name
brain expressed X-linked 4
Gene Aliases
BEXL1
UniGene
Hs.184736
Gene Chromosome Location
Chr.X: 103215092-103217200 on Build GRCh38
Transcripts
NM_001080425.3 NM_001127688.2

SNP Detail

SNP ID Chromosome Location
rs773879410 Chr.X: 103216302-103216302 on Build GRCh38
rs750194488 Chr.X: 103216210-103216210 on Build GRCh38
rs201463444 Chr.X: 103216206-103216206 on Build GRCh38
rs778341539 Chr.X: 103216234-103216234 on Build GRCh38
rs752813806 Chr.X: 103216365-103216365 on Build GRCh38
rs767426858 Chr.X: 103216308-103216308 on Build GRCh38
rs943595788 Chr.X: 103216144-103216144 on Build GRCh38
rs754041965 Chr.X: 103216350-103216350 on Build GRCh38
rs760463621 Chr.X: 103216314-103216314 on Build GRCh38
rs931646299 Chr.X: 103216165-103216165 on Build GRCh38
rs772687582 Chr.X: 103216233-103216233 on Build GRCh38
rs3179998 Chr.X: 103216300-103216300 on Build GRCh38
rs751472806 Chr.X: 103216186-103216186 on Build GRCh38
rs149236674 Chr.X: 103216362-103216362 on Build GRCh38
rs62623366 Chr.X: 103216307-103216307 on Build GRCh38
rs764419031 Chr.X: 103216288-103216288 on Build GRCh38
rs372941608 Chr.X: 103216279-103216279 on Build GRCh38
rs200516702 Chr.X: 103216243-103216243 on Build GRCh38
rs775129946 Chr.X: 103216212-103216212 on Build GRCh38
rs150891402 Chr.X: 103216163-103216163 on Build GRCh38
rs936504299 Chr.X: 103216298-103216298 on Build GRCh38
rs775018543 Chr.X: 103216262-103216262 on Build GRCh38
rs779013664 Chr.X: 103216228-103216228 on Build GRCh38
rs747374478 Chr.X: 103216239-103216239 on Build GRCh38
rs780131469 Chr.X: 103216214-103216214 on Build GRCh38
rs370714145 Chr.X: 103216185-103216185 on Build GRCh38
rs761698011 Chr.X: 103216202-103216202 on Build GRCh38
rs755040139 Chr.X: 103216221-103216221 on Build GRCh38
rs892386748 Chr.X: 103216315-103216315 on Build GRCh38
rs750282321 Chr.X: 103216310-103216310 on Build GRCh38
rs185873042 Chr.X: 103216339-103216339 on Build GRCh38
rs754020950 Chr.X: 103216217-103216217 on Build GRCh38
rs748708849 Chr.X: 103216231-103216231 on Build GRCh38
rs755198979 Chr.X: 103216361-103216361 on Build GRCh38
rs376522320 Chr.X: 103216151-103216151 on Build GRCh38

View Full Product Details