Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29071812-29072079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf71
Entrez Gene ID
388939
Gene Name
chromosome 2 open reading frame 71
Gene Aliases
RP54
UniGene
Hs.354243
Gene Chromosome Location
Chr.2: 29061690-29074261 on Build GRCh38
Transcripts
NM_001029883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334455 c.2356G>T p.A786S Chr.2: 29071906-29071906 on Build GRCh38
3580946 c.2243G>A p.G748E Chr.2: 29072019-29072019 on Build GRCh38
6880410 c.2235G>A p.R745R Chr.2: 29072027-29072027 on Build GRCh38
5558257 c.2194A>C p.K732Q Chr.2: 29072068-29072068 on Build GRCh38
1198732 c.2351C>A p.S784Y Chr.2: 29071911-29071911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865943606 Chr.2: 29071957-29071957 on Build GRCh38
rs886055921 Chr.2: 29071975-29071975 on Build GRCh38
rs762281847 Chr.2: 29071960-29071961 on Build GRCh38
rs1002848166 Chr.2: 29071999-29071999 on Build GRCh38
rs949611430 Chr.2: 29071881-29071881 on Build GRCh38
rs769809028 Chr.2: 29071901-29071901 on Build GRCh38
rs373055833 Chr.2: 29072039-29072039 on Build GRCh38
rs34522598 Chr.2: 29072021-29072022 on Build GRCh38
rs919932708 Chr.2: 29072014-29072014 on Build GRCh38
rs371979312 Chr.2: 29072005-29072005 on Build GRCh38
rs751929049 Chr.2: 29071884-29071884 on Build GRCh38
rs757721465 Chr.2: 29071931-29071931 on Build GRCh38
rs776283046 Chr.2: 29071978-29071978 on Build GRCh38
rs780585710 Chr.2: 29071869-29071869 on Build GRCh38
rs773043850 Chr.2: 29072002-29072002 on Build GRCh38
rs768595767 Chr.2: 29071973-29071973 on Build GRCh38
rs772053407 Chr.2: 29071918-29071918 on Build GRCh38
rs200963687 Chr.2: 29071893-29071893 on Build GRCh38
rs750088901 Chr.2: 29071937-29071937 on Build GRCh38
rs761471052 Chr.2: 29071984-29071984 on Build GRCh38
rs141559332 Chr.2: 29071914-29071914 on Build GRCh38
rs746938428 Chr.2: 29071972-29071972 on Build GRCh38
rs371909714 Chr.2: 29071838-29071838 on Build GRCh38
rs982843986 Chr.2: 29072017-29072017 on Build GRCh38
rs756596110 Chr.2: 29071951-29071951 on Build GRCh38
rs757393716 Chr.2: 29072026-29072026 on Build GRCh38
rs763558017 Chr.2: 29071926-29071926 on Build GRCh38
rs779742136 Chr.2: 29072041-29072041 on Build GRCh38
rs867892364 Chr.2: 29071845-29071846 on Build GRCh38
rs375644198 Chr.2: 29072006-29072006 on Build GRCh38
rs886044002 Chr.2: 29072034-29072035 on Build GRCh38
rs777887655 Chr.2: 29071904-29071904 on Build GRCh38
rs779162226 Chr.2: 29071965-29071965 on Build GRCh38
rs369823170 Chr.2: 29072034-29072034 on Build GRCh38
rs771841315 Chr.2: 29071923-29071923 on Build GRCh38
rs185637100 Chr.2: 29071989-29071989 on Build GRCh38
rs181545106 Chr.2: 29071916-29071916 on Build GRCh38
rs757483315 Chr.2: 29071964-29071964 on Build GRCh38
rs868665415 Chr.2: 29071921-29071921 on Build GRCh38
rs764602216 Chr.2: 29072008-29072008 on Build GRCh38
rs907204029 Chr.2: 29071999-29071999 on Build GRCh38
rs189042259 Chr.2: 29071844-29071844 on Build GRCh38
rs753194513 Chr.2: 29071948-29071948 on Build GRCh38
rs200759534 Chr.2: 29071953-29071953 on Build GRCh38
rs761585411 Chr.2: 29071932-29071932 on Build GRCh38
rs17744093 Chr.2: 29071888-29071888 on Build GRCh38
rs150881512 Chr.2: 29071971-29071971 on Build GRCh38
rs753470385 Chr.2: 29071853-29071853 on Build GRCh38
rs751935156 Chr.2: 29071925-29071925 on Build GRCh38
rs375771666 Chr.2: 29071856-29071856 on Build GRCh38
rs866399704 Chr.2: 29071844-29071845 on Build GRCh38
rs1041787727 Chr.2: 29071992-29071992 on Build GRCh38
rs746085242 Chr.2: 29071969-29071969 on Build GRCh38
rs781609460 Chr.2: 29071891-29071891 on Build GRCh38
rs764928525 Chr.2: 29071934-29071934 on Build GRCh38
rs781754063 Chr.2: 29072028-29072028 on Build GRCh38
rs767896465 Chr.2: 29071947-29071947 on Build GRCh38
rs951445929 Chr.2: 29072016-29072016 on Build GRCh38
rs752263111 Chr.2: 29071839-29071839 on Build GRCh38
rs758592526 Chr.2: 29072040-29072040 on Build GRCh38
rs760160585 Chr.2: 29071842-29071842 on Build GRCh38
rs749301631 Chr.2: 29071962-29071962 on Build GRCh38
rs762676298 Chr.2: 29072003-29072003 on Build GRCh38
rs749536412 Chr.2: 29071913-29071913 on Build GRCh38
rs376533270 Chr.2: 29071939-29071939 on Build GRCh38
rs541711950 Chr.2: 29071970-29071970 on Build GRCh38
rs754327204 Chr.2: 29072020-29072020 on Build GRCh38

View Full Product Details