Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94761505-94761762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L4
Entrez Gene ID
286148
Gene Name
dpy-19 like 4 (C. elegans)
Gene Aliases
-
UniGene
Hs.567828
Gene Chromosome Location
Chr.8: 94719708-94793848 on Build GRCh38
Transcripts
NM_181787.2 XM_005250894.4 XM_005250895.3 XM_011516983.2 XM_011516984.2 XM_017013351.1 XM_017013352.1 XM_017013353.1 XM_017013354.1 XM_017013355.1 XM_017013356.1 XM_017013357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
486769 c.791A>G p.E264G Chr.8: 94761755-94761755 on Build GRCh38
6181720 c.736-2A>T p.? Chr.8: 94761698-94761698 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866047601 Chr.8: 94761695-94761695 on Build GRCh38
rs145462618 Chr.8: 94761730-94761730 on Build GRCh38
rs529179350 Chr.8: 94761668-94761668 on Build GRCh38
rs138678169 Chr.8: 94761659-94761660 on Build GRCh38
rs1024197587 Chr.8: 94761713-94761713 on Build GRCh38
rs764019092 Chr.8: 94761715-94761715 on Build GRCh38
rs536070654 Chr.8: 94761593-94761593 on Build GRCh38
rs943382746 Chr.8: 94761540-94761540 on Build GRCh38
rs377151287 Chr.8: 94761651-94761651 on Build GRCh38
rs555635846 Chr.8: 94761654-94761654 on Build GRCh38
rs752984771 Chr.8: 94761689-94761689 on Build GRCh38
rs760766744 Chr.8: 94761698-94761698 on Build GRCh38
rs902069319 Chr.8: 94761602-94761602 on Build GRCh38
rs888209722 Chr.8: 94761531-94761531 on Build GRCh38
rs759661427 Chr.8: 94761681-94761681 on Build GRCh38
rs1034059277 Chr.8: 94761625-94761625 on Build GRCh38
rs774477610 Chr.8: 94761665-94761665 on Build GRCh38
rs993628850 Chr.8: 94761605-94761605 on Build GRCh38
rs1014027252 Chr.8: 94761696-94761696 on Build GRCh38
rs752848278 Chr.8: 94761572-94761572 on Build GRCh38
rs1041061709 Chr.8: 94761579-94761579 on Build GRCh38
rs767438570 Chr.8: 94761688-94761688 on Build GRCh38
rs958563200 Chr.8: 94761641-94761641 on Build GRCh38
rs1005673907 Chr.8: 94761606-94761606 on Build GRCh38
rs1043807938 Chr.8: 94761530-94761530 on Build GRCh38

View Full Product Details