Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15008291-15008550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB2
Entrez Gene ID
132864
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
CPE-BP2, CPEB-2, hCPEB-2
UniGene
Hs.656937
Gene Chromosome Location
Chr.4: 15002451-15070153 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1 XM_017007733.1 XM_017007734.1 NR_125911.1

Gene Symbol
LOC101929095
Entrez Gene ID
101929095
Gene Name
uncharacterized LOC101929095
Gene Aliases
-
UniGene
Hs.656908
Gene Chromosome Location
Chr.4: 15004942-15427914 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1 XM_017007733.1 XM_017007734.1 NR_125911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1685137 c.1961A>G p.Q654R Chr.4: 15008354-15008354 on Build GRCh38
6400799 c.674C>A p.S225* Chr.4: 15008378-15008378 on Build GRCh38
6634516 c.693C>A p.C231* Chr.4: 15008397-15008397 on Build GRCh38
6371630 c.2014G>A p.G672R Chr.4: 15008407-15008407 on Build GRCh38
6634515 c.2004C>A p.C668* Chr.4: 15008397-15008397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748231286 Chr.4: 15008357-15008357 on Build GRCh38
rs943285004 Chr.4: 15008502-15008502 on Build GRCh38
rs768939954 Chr.4: 15008326-15008326 on Build GRCh38
rs780150401 Chr.4: 15008460-15008460 on Build GRCh38
rs556836059 Chr.4: 15008448-15008448 on Build GRCh38
rs776924854 Chr.4: 15008388-15008388 on Build GRCh38
rs1031712574 Chr.4: 15008482-15008482 on Build GRCh38
rs572067309 Chr.4: 15008510-15008510 on Build GRCh38
rs369684904 Chr.4: 15008461-15008461 on Build GRCh38
rs998333856 Chr.4: 15008395-15008395 on Build GRCh38
rs772097287 Chr.4: 15008362-15008362 on Build GRCh38
rs536403362 Chr.4: 15008446-15008446 on Build GRCh38
rs565976876 Chr.4: 15008513-15008513 on Build GRCh38
rs960947184 Chr.4: 15008499-15008499 on Build GRCh38
rs751209038 Chr.4: 15008440-15008440 on Build GRCh38
rs763694978 Chr.4: 15008411-15008411 on Build GRCh38
rs773760698 Chr.4: 15008369-15008369 on Build GRCh38
rs779176137 Chr.4: 15008470-15008470 on Build GRCh38
rs759918003 Chr.4: 15008405-15008405 on Build GRCh38
rs774470310 Chr.4: 15008344-15008344 on Build GRCh38
rs756045027 Chr.4: 15008455-15008455 on Build GRCh38
rs762928600 Chr.4: 15008493-15008493 on Build GRCh38
rs558293988 Chr.4: 15008494-15008494 on Build GRCh38
rs376443106 Chr.4: 15008385-15008385 on Build GRCh38
rs539300300 Chr.4: 15008492-15008492 on Build GRCh38
rs761153360 Chr.4: 15008380-15008380 on Build GRCh38
rs892544070 Chr.4: 15008419-15008419 on Build GRCh38
rs756768908 Chr.4: 15008442-15008442 on Build GRCh38
rs1013143521 Chr.4: 15008406-15008406 on Build GRCh38
rs75021526 Chr.4: 15008489-15008489 on Build GRCh38
rs1011002516 Chr.4: 15008450-15008450 on Build GRCh38

View Full Product Details