Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56491382-56491653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT7
Entrez Gene ID
140687
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases
C20orf105, dJ1153D9.2, gcnt
UniGene
Hs.655916
Gene Chromosome Location
Chr.20: 56491492-56525925 on Build GRCh38
Transcripts
NM_080615.1 NM_001283035.1 NM_001283036.1 NM_001283037.1 NM_016407.4 XM_017027872.1

Gene Symbol
RTFDC1
Entrez Gene ID
51507
Gene Name
replication termination factor 2 domain containing 1
Gene Aliases
C20orf43, CDAO5, HSPC164, SHUJUN-3
UniGene
Hs.517134
Gene Chromosome Location
Chr.20: 56468585-56518886 on Build GRCh38
Transcripts
NM_080615.1 NM_001283035.1 NM_001283036.1 NM_001283037.1 NM_016407.4 XM_017027872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1592678 c.901C>A p.H301N Chr.20: 56491583-56491583 on Build GRCh38
4972624 c.879C>G p.H293Q Chr.20: 56491605-56491605 on Build GRCh38
4001961 c.907A>C p.N303H Chr.20: 56491577-56491577 on Build GRCh38
5783759 c.914G>C p.R305T Chr.20: 56491570-56491570 on Build GRCh38
3933884 c.831G>A p.R277R Chr.20: 56491653-56491653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766895347 Chr.20: 56491563-56491563 on Build GRCh38
rs929581532 Chr.20: 56491535-56491535 on Build GRCh38
rs904292740 Chr.20: 56491562-56491562 on Build GRCh38
rs183774505 Chr.20: 56491559-56491559 on Build GRCh38
rs113542963 Chr.20: 56491443-56491443 on Build GRCh38
rs765004088 Chr.20: 56491428-56491428 on Build GRCh38
rs562870802 Chr.20: 56491471-56491471 on Build GRCh38
rs749406886 Chr.20: 56491601-56491601 on Build GRCh38
rs777539134 Chr.20: 56491518-56491548 on Build GRCh38
rs1052669455 Chr.20: 56491440-56491440 on Build GRCh38
rs188301833 Chr.20: 56491561-56491561 on Build GRCh38
rs780187050 Chr.20: 56491614-56491614 on Build GRCh38
rs910639673 Chr.20: 56491470-56491470 on Build GRCh38
rs3746627 Chr.20: 56491577-56491577 on Build GRCh38
rs56066546 Chr.20: 56491475-56491475 on Build GRCh38
rs768329860 Chr.20: 56491533-56491533 on Build GRCh38
rs780643587 Chr.20: 56491574-56491574 on Build GRCh38
rs764784554 Chr.20: 56491557-56491557 on Build GRCh38
rs758359093 Chr.20: 56491610-56491610 on Build GRCh38
rs998141826 Chr.20: 56491576-56491576 on Build GRCh38
rs372122252 Chr.20: 56491546-56491546 on Build GRCh38
rs755340714 Chr.20: 56491573-56491573 on Build GRCh38
rs752922889 Chr.20: 56491581-56491581 on Build GRCh38
rs985662565 Chr.20: 56491431-56491431 on Build GRCh38
rs138510750 Chr.20: 56491556-56491556 on Build GRCh38
rs1000326944 Chr.20: 56491566-56491566 on Build GRCh38
rs916582361 Chr.20: 56491597-56491597 on Build GRCh38
rs756375751 Chr.20: 56491589-56491589 on Build GRCh38
rs543725406 Chr.20: 56491615-56491615 on Build GRCh38
rs891386401 Chr.20: 56491468-56491468 on Build GRCh38
rs191665128 Chr.20: 56491498-56491498 on Build GRCh38
rs141674963 Chr.20: 56491567-56491567 on Build GRCh38
rs777937654 Chr.20: 56491592-56491592 on Build GRCh38

View Full Product Details