Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102929796-102930031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN
Entrez Gene ID
81693
Gene Name
amnion associated transmembrane protein
Gene Aliases
PRO1028, amnionless
UniGene
Hs.534494
Gene Chromosome Location
Chr.14: 102922430-102930842 on Build GRCh38
Transcripts
NM_030943.3 XM_011537202.2 XM_011537203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1368376 c.851A>G p.Y284C Chr.14: 102929931-102929931 on Build GRCh38
4885853 c.870C>A p.A290A Chr.14: 102929950-102929950 on Build GRCh38
3689946 c.944G>T p.G315V Chr.14: 102930024-102930024 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs995278041 Chr.14: 102929850-102929850 on Build GRCh38
rs866436007 Chr.14: 102929994-102929994 on Build GRCh38
rs760766577 Chr.14: 102929998-102929998 on Build GRCh38
rs779650808 Chr.14: 102929975-102929975 on Build GRCh38
rs373382273 Chr.14: 102929989-102929989 on Build GRCh38
rs1031050005 Chr.14: 102929885-102929886 on Build GRCh38
rs201788173 Chr.14: 102930008-102930008 on Build GRCh38
rs959226628 Chr.14: 102929868-102929868 on Build GRCh38
rs746596986 Chr.14: 102929965-102929965 on Build GRCh38
rs779550596 Chr.14: 102929913-102929913 on Build GRCh38
rs766601907 Chr.14: 102929883-102929883 on Build GRCh38
rs557645687 Chr.14: 102929873-102929873 on Build GRCh38
rs866773035 Chr.14: 102929964-102929964 on Build GRCh38
rs145519816 Chr.14: 102929921-102929921 on Build GRCh38
rs777586051 Chr.14: 102929962-102929962 on Build GRCh38
rs769673963 Chr.14: 102929982-102929982 on Build GRCh38
rs745651516 Chr.14: 102929978-102929978 on Build GRCh38
rs990743801 Chr.14: 102929886-102929886 on Build GRCh38
rs963445170 Chr.14: 102929846-102929846 on Build GRCh38
rs899688048 Chr.14: 102929823-102929823 on Build GRCh38
rs958270070 Chr.14: 102929898-102929898 on Build GRCh38
rs772080788 Chr.14: 102929990-102929990 on Build GRCh38
rs770793845 Chr.14: 102929970-102929970 on Build GRCh38
rs1014976990 Chr.14: 102929838-102929838 on Build GRCh38
rs1030387713 Chr.14: 102929852-102929852 on Build GRCh38
rs773444418 Chr.14: 102929995-102929995 on Build GRCh38
rs1005297327 Chr.14: 102929832-102929832 on Build GRCh38
rs755488212 Chr.14: 102929909-102929909 on Build GRCh38
rs1026756993 Chr.14: 102929856-102929856 on Build GRCh38
rs969552874 Chr.14: 102929923-102929923 on Build GRCh38
rs866260453 Chr.14: 102929963-102929963 on Build GRCh38
rs369865835 Chr.14: 102929935-102929935 on Build GRCh38
rs867728933 Chr.14: 102929949-102929949 on Build GRCh38
rs890460212 Chr.14: 102929830-102929830 on Build GRCh38
rs927694676 Chr.14: 102929958-102929958 on Build GRCh38
rs761114053 Chr.14: 102929877-102929877 on Build GRCh38
rs754398846 Chr.14: 102929885-102929885 on Build GRCh38
rs775398085 Chr.14: 102929984-102929984 on Build GRCh38

View Full Product Details