Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19483811-19484052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC45
Entrez Gene ID
8318
Gene Name
cell division cycle 45
Gene Aliases
CDC45L, CDC45L2, PORC-PI-1
UniGene
Hs.474217
Gene Chromosome Location
Chr.22: 19479278-19520612 on Build GRCh38
Transcripts
NM_001178010.2 NM_001178011.2 NM_003504.4 XM_005261285.2 XM_005261286.3 XM_011530415.1 XM_011530416.1 XM_011530417.2 XM_011530418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7360415 c.449G>A p.G150E Chr.22: 19483968-19483968 on Build GRCh38
5907778 c.458C>T p.P153L Chr.22: 19483977-19483977 on Build GRCh38
5858908 c.470G>A p.R157H Chr.22: 19483989-19483989 on Build GRCh38
4937798 c.395A>G p.D132G Chr.22: 19483914-19483914 on Build GRCh38
6624455 c.476G>A p.R159Q Chr.22: 19483995-19483995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747388741 Chr.22: 19483981-19483981 on Build GRCh38
rs747335297 Chr.22: 19483903-19483903 on Build GRCh38
rs772870688 Chr.22: 19484022-19484022 on Build GRCh38
rs746926103 Chr.22: 19483932-19483932 on Build GRCh38
rs9606030 Chr.22: 19483983-19483983 on Build GRCh38
rs758094694 Chr.22: 19483963-19483963 on Build GRCh38
rs777216317 Chr.22: 19483906-19483906 on Build GRCh38
rs145708400 Chr.22: 19483962-19483962 on Build GRCh38
rs369176967 Chr.22: 19483904-19483904 on Build GRCh38
rs112518152 Chr.22: 19484004-19484004 on Build GRCh38
rs575366448 Chr.22: 19483868-19483868 on Build GRCh38
rs759588471 Chr.22: 19483909-19483909 on Build GRCh38
rs61752240 Chr.22: 19483937-19483937 on Build GRCh38
rs967159298 Chr.22: 19484016-19484016 on Build GRCh38
rs202172934 Chr.22: 19484005-19484005 on Build GRCh38
rs750251566 Chr.22: 19483930-19483930 on Build GRCh38
rs914630577 Chr.22: 19484029-19484029 on Build GRCh38
rs957839823 Chr.22: 19483989-19483989 on Build GRCh38
rs140290119 Chr.22: 19483947-19483947 on Build GRCh38
rs373418450 Chr.22: 19483994-19483994 on Build GRCh38
rs763210706 Chr.22: 19483915-19483915 on Build GRCh38
rs758575467 Chr.22: 19483874-19483874 on Build GRCh38
rs764249763 Chr.22: 19483921-19483921 on Build GRCh38
rs780246597 Chr.22: 19484017-19484018 on Build GRCh38
rs149459036 Chr.22: 19483884-19483884 on Build GRCh38
rs775615531 Chr.22: 19483914-19483914 on Build GRCh38
rs546620592 Chr.22: 19484010-19484010 on Build GRCh38
rs377186449 Chr.22: 19483955-19483955 on Build GRCh38
rs764306890 Chr.22: 19484014-19484014 on Build GRCh38
rs766316844 Chr.22: 19483938-19483938 on Build GRCh38
rs945389583 Chr.22: 19483879-19483879 on Build GRCh38
rs200034634 Chr.22: 19483891-19483891 on Build GRCh38
rs774498452 Chr.22: 19484019-19484019 on Build GRCh38
rs753801276 Chr.22: 19483943-19483943 on Build GRCh38
rs769448408 Chr.22: 19483995-19483995 on Build GRCh38
rs1025020064 Chr.22: 19483964-19483964 on Build GRCh38
rs765355152 Chr.22: 19483913-19483913 on Build GRCh38
rs534789113 Chr.22: 19483975-19483975 on Build GRCh38
rs778459455 Chr.22: 19483960-19483960 on Build GRCh38
rs756450477 Chr.22: 19483931-19483933 on Build GRCh38
rs754454198 Chr.22: 19483883-19483883 on Build GRCh38
rs779928712 Chr.22: 19483862-19483862 on Build GRCh38
rs756069986 Chr.22: 19483842-19483842 on Build GRCh38
rs1032293178 Chr.22: 19483987-19483987 on Build GRCh38
rs554571750 Chr.22: 19483892-19483892 on Build GRCh38
rs540217942 Chr.22: 19483988-19483988 on Build GRCh38
rs866965652 Chr.22: 19483911-19483911 on Build GRCh38

View Full Product Details