Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23910202-23910475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf58
Entrez Gene ID
254158
Gene Name
chromosome X open reading frame 58
Gene Aliases
-
UniGene
Hs.351265
Gene Chromosome Location
Chr.X: 23907932-23939520 on Build GRCh38
Transcripts
NM_001169574.1 NM_152761.2 XM_011545473.2

SNP Detail

SNP ID Chromosome Location
rs769461268 Chr.X: 23910396-23910396 on Build GRCh38
rs776863884 Chr.X: 23910422-23910422 on Build GRCh38
rs755980339 Chr.X: 23910419-23910420 on Build GRCh38
rs2707164 Chr.X: 23910372-23910372 on Build GRCh38
rs750942077 Chr.X: 23910353-23910353 on Build GRCh38
rs763187649 Chr.X: 23910286-23910286 on Build GRCh38
rs375970076 Chr.X: 23910272-23910272 on Build GRCh38
rs773222705 Chr.X: 23910273-23910273 on Build GRCh38
rs149951169 Chr.X: 23910364-23910364 on Build GRCh38
rs761448822 Chr.X: 23910271-23910271 on Build GRCh38
rs1055514208 Chr.X: 23910412-23910413 on Build GRCh38
rs770832471 Chr.X: 23910416-23910416 on Build GRCh38
rs749264016 Chr.X: 23910413-23910413 on Build GRCh38
rs745815273 Chr.X: 23910283-23910283 on Build GRCh38
rs199875318 Chr.X: 23910417-23910417 on Build GRCh38
rs144666555 Chr.X: 23910386-23910386 on Build GRCh38
rs752366803 Chr.X: 23910242-23910242 on Build GRCh38
rs369099467 Chr.X: 23910291-23910291 on Build GRCh38
rs754032781 Chr.X: 23910334-23910334 on Build GRCh38
rs141234868 Chr.X: 23910309-23910309 on Build GRCh38
rs78304044 Chr.X: 23910418-23910418 on Build GRCh38
rs772530059 Chr.X: 23910399-23910399 on Build GRCh38
rs764370346 Chr.X: 23910327-23910327 on Build GRCh38
rs755363601 Chr.X: 23910378-23910378 on Build GRCh38
rs1052598729 Chr.X: 23910241-23910241 on Build GRCh38
rs761953508 Chr.X: 23910446-23910446 on Build GRCh38
rs767281926 Chr.X: 23910419-23910419 on Build GRCh38
rs756088137 Chr.X: 23910303-23910303 on Build GRCh38
rs1039467650 Chr.X: 23910323-23910323 on Build GRCh38
rs757379317 Chr.X: 23910335-23910335 on Build GRCh38
rs139980314 Chr.X: 23910322-23910322 on Build GRCh38
rs147890788 Chr.X: 23910387-23910387 on Build GRCh38
rs779844812 Chr.X: 23910447-23910447 on Build GRCh38
rs765187437 Chr.X: 23910337-23910337 on Build GRCh38
rs113507987 Chr.X: 23910365-23910365 on Build GRCh38
rs980768078 Chr.X: 23910439-23910439 on Build GRCh38

View Full Product Details