Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 147981224-147981493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMR1NB
Entrez Gene ID
158521
Gene Name
fragile X mental retardation 1 neighbor
Gene Aliases
CT37, NY-SAR-35, NYSAR35
UniGene
Hs.128580
Gene Chromosome Location
Chr.X: 147981329-148026667 on Build GRCh38
Transcripts
NM_152578.2

SNP Detail

SNP ID Chromosome Location
rs782236986 Chr.X: 147981374-147981375 on Build GRCh38
rs782256745 Chr.X: 147981440-147981440 on Build GRCh38
rs782209231 Chr.X: 147981381-147981381 on Build GRCh38
rs782415527 Chr.X: 147981388-147981388 on Build GRCh38
rs782631524 Chr.X: 147981395-147981395 on Build GRCh38
rs782564160 Chr.X: 147981434-147981434 on Build GRCh38
rs1014370779 Chr.X: 147981301-147981301 on Build GRCh38
rs921733761 Chr.X: 147981392-147981392 on Build GRCh38
rs782468233 Chr.X: 147981425-147981425 on Build GRCh38
rs1052101549 Chr.X: 147981279-147981279 on Build GRCh38
rs782132283 Chr.X: 147981417-147981417 on Build GRCh38
rs782656279 Chr.X: 147981437-147981437 on Build GRCh38
rs906100027 Chr.X: 147981324-147981324 on Build GRCh38
rs372262278 Chr.X: 147981431-147981431 on Build GRCh38
rs1027673330 Chr.X: 147981345-147981345 on Build GRCh38
rs191218185 Chr.X: 147981377-147981377 on Build GRCh38
rs782088001 Chr.X: 147981357-147981357 on Build GRCh38
rs370614051 Chr.X: 147981354-147981354 on Build GRCh38
rs200471047 Chr.X: 147981461-147981461 on Build GRCh38
rs924918139 Chr.X: 147981269-147981269 on Build GRCh38
rs782401588 Chr.X: 147981467-147981467 on Build GRCh38
rs200555900 Chr.X: 147981382-147981382 on Build GRCh38
rs782366161 Chr.X: 147981450-147981450 on Build GRCh38
rs201101798 Chr.X: 147981426-147981426 on Build GRCh38
rs782345347 Chr.X: 147981394-147981394 on Build GRCh38
rs202129849 Chr.X: 147981376-147981376 on Build GRCh38
rs782781440 Chr.X: 147981418-147981418 on Build GRCh38
rs782722857 Chr.X: 147981302-147981302 on Build GRCh38
rs370744869 Chr.X: 147981380-147981380 on Build GRCh38
rs1046426071 Chr.X: 147981323-147981323 on Build GRCh38
rs781996475 Chr.X: 147981389-147981389 on Build GRCh38
rs368451527 Chr.X: 147981385-147981385 on Build GRCh38
rs1043412995 Chr.X: 147981266-147981266 on Build GRCh38
rs781876920 Chr.X: 147981378-147981378 on Build GRCh38
rs1002163349 Chr.X: 147981334-147981334 on Build GRCh38
rs782201815 Chr.X: 147981452-147981452 on Build GRCh38
rs782606068 Chr.X: 147981360-147981361 on Build GRCh38
rs935086218 Chr.X: 147981275-147981275 on Build GRCh38
rs892000792 Chr.X: 147981296-147981296 on Build GRCh38
rs182787302 Chr.X: 147981390-147981390 on Build GRCh38
rs782607537 Chr.X: 147981451-147981451 on Build GRCh38
rs781834891 Chr.X: 147981420-147981420 on Build GRCh38
rs375204379 Chr.X: 147981460-147981460 on Build GRCh38
rs782329475 Chr.X: 147981413-147981413 on Build GRCh38
rs782570167 Chr.X: 147981373-147981373 on Build GRCh38
rs782730280 Chr.X: 147981358-147981358 on Build GRCh38
rs187764243 Chr.X: 147981397-147981397 on Build GRCh38
rs372849176 Chr.X: 147981359-147981359 on Build GRCh38
rs782473536 Chr.X: 147981358-147981359 on Build GRCh38

View Full Product Details