Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127737420-127737667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI2
Entrez Gene ID
399823
Gene Name
forkhead box I2
Gene Aliases
-
UniGene
Hs.449674
Gene Chromosome Location
Chr.10: 127737274-127741186 on Build GRCh38
Transcripts
NM_207426.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940032 c.386A>C p.Y129S Chr.10: 127737659-127737659 on Build GRCh38
7289619 c.309G>A p.P103P Chr.10: 127737582-127737582 on Build GRCh38
4797805 c.351C>T p.S117S Chr.10: 127737624-127737624 on Build GRCh38
6431195 c.334G>T p.A112S Chr.10: 127737607-127737607 on Build GRCh38
6397742 c.248G>A p.G83D Chr.10: 127737521-127737521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575900552 Chr.10: 127737470-127737470 on Build GRCh38
rs921718115 Chr.10: 127737532-127737532 on Build GRCh38
rs780937943 Chr.10: 127737629-127737629 on Build GRCh38
rs778494055 Chr.10: 127737575-127737575 on Build GRCh38
rs1048806989 Chr.10: 127737574-127737574 on Build GRCh38
rs533064876 Chr.10: 127737627-127737627 on Build GRCh38
rs896387507 Chr.10: 127737625-127737625 on Build GRCh38
rs931720720 Chr.10: 127737533-127737533 on Build GRCh38
rs118034115 Chr.10: 127737455-127737455 on Build GRCh38
rs370357053 Chr.10: 127737632-127737632 on Build GRCh38
rs979447024 Chr.10: 127737464-127737464 on Build GRCh38
rs750778638 Chr.10: 127737509-127737509 on Build GRCh38
rs909280643 Chr.10: 127737446-127737446 on Build GRCh38
rs557531167 Chr.10: 127737462-127737462 on Build GRCh38
rs992557851 Chr.10: 127737445-127737445 on Build GRCh38
rs970796351 Chr.10: 127737571-127737571 on Build GRCh38
rs912787136 Chr.10: 127737501-127737501 on Build GRCh38
rs752345259 Chr.10: 127737597-127737597 on Build GRCh38
rs573556148 Chr.10: 127737531-127737531 on Build GRCh38
rs765560530 Chr.10: 127737484-127737484 on Build GRCh38
rs969327418 Chr.10: 127737636-127737636 on Build GRCh38
rs1009695229 Chr.10: 127737549-127737549 on Build GRCh38
rs1007713632 Chr.10: 127737522-127737522 on Build GRCh38
rs562046749 Chr.10: 127737481-127737481 on Build GRCh38
rs866944235 Chr.10: 127737491-127737491 on Build GRCh38
rs762112354 Chr.10: 127737637-127737637 on Build GRCh38
rs373919174 Chr.10: 127737610-127737610 on Build GRCh38
rs543325801 Chr.10: 127737476-127737476 on Build GRCh38
rs1036311998 Chr.10: 127737611-127737611 on Build GRCh38
rs113343672 Chr.10: 127737543-127737543 on Build GRCh38
rs377396967 Chr.10: 127737635-127737635 on Build GRCh38
rs956841998 Chr.10: 127737485-127737485 on Build GRCh38
rs1020704788 Chr.10: 127737552-127737552 on Build GRCh38
rs758107251 Chr.10: 127737600-127737600 on Build GRCh38
rs541064042 Chr.10: 127737578-127737578 on Build GRCh38

View Full Product Details