Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51299494-51299767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN2
Entrez Gene ID
51411
Gene Name
bridging integrator 2
Gene Aliases
BRAP-1
UniGene
Hs.14770
Gene Chromosome Location
Chr.12: 51281038-51324680 on Build GRCh38
Transcripts
NM_001290007.1 NM_001290008.1 NM_001290009.1 NM_016293.3 XM_005268957.3 XM_005268958.3 XM_017019422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4042677 c.462C>T p.H154H Chr.12: 51299661-51299661 on Build GRCh38
1492927 c.516+2T>A p.? Chr.12: 51299605-51299605 on Build GRCh38
6045768 c.457C>T p.R153* Chr.12: 51299666-51299666 on Build GRCh38
6025313 c.440T>C p.V147A Chr.12: 51299683-51299683 on Build GRCh38
4042678 c.430C>T p.R144W Chr.12: 51299693-51299693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768632661 Chr.12: 51299563-51299563 on Build GRCh38
rs910475849 Chr.12: 51299557-51299557 on Build GRCh38
rs770156666 Chr.12: 51299590-51299590 on Build GRCh38
rs575604709 Chr.12: 51299567-51299567 on Build GRCh38
rs768014177 Chr.12: 51299738-51299738 on Build GRCh38
rs897449285 Chr.12: 51299553-51299553 on Build GRCh38
rs986013549 Chr.12: 51299551-51299551 on Build GRCh38
rs777651541 Chr.12: 51299696-51299696 on Build GRCh38
rs150740618 Chr.12: 51299631-51299631 on Build GRCh38
rs772029129 Chr.12: 51299587-51299587 on Build GRCh38
rs969445320 Chr.12: 51299693-51299693 on Build GRCh38
rs758015253 Chr.12: 51299586-51299588 on Build GRCh38
rs753374931 Chr.12: 51299666-51299666 on Build GRCh38
rs763013855 Chr.12: 51299605-51299605 on Build GRCh38
rs762135863 Chr.12: 51299624-51299624 on Build GRCh38
rs1032850813 Chr.12: 51299740-51299740 on Build GRCh38
rs762262605 Chr.12: 51299729-51299729 on Build GRCh38
rs770052008 Chr.12: 51299698-51299698 on Build GRCh38
rs139065319 Chr.12: 51299665-51299665 on Build GRCh38
rs192943143 Chr.12: 51299638-51299638 on Build GRCh38
rs929110424 Chr.12: 51299594-51299594 on Build GRCh38
rs749365314 Chr.12: 51299701-51299701 on Build GRCh38
rs200130190 Chr.12: 51299588-51299588 on Build GRCh38
rs199541031 Chr.12: 51299548-51299548 on Build GRCh38
rs779557037 Chr.12: 51299560-51299560 on Build GRCh38
rs774523510 Chr.12: 51299710-51299710 on Build GRCh38
rs774401820 Chr.12: 51299669-51299669 on Build GRCh38
rs766068626 Chr.12: 51299614-51299615 on Build GRCh38
rs767830479 Chr.12: 51299626-51299626 on Build GRCh38
rs559036732 Chr.12: 51299555-51299555 on Build GRCh38
rs201767225 Chr.12: 51299628-51299628 on Build GRCh38
rs375843733 Chr.12: 51299742-51299742 on Build GRCh38
rs78442656 Chr.12: 51299685-51299685 on Build GRCh38
rs374011518 Chr.12: 51299699-51299699 on Build GRCh38
rs202182807 Chr.12: 51299600-51299600 on Build GRCh38
rs748057174 Chr.12: 51299686-51299686 on Build GRCh38
rs577030938 Chr.12: 51299645-51299645 on Build GRCh38
rs755797466 Chr.12: 51299556-51299556 on Build GRCh38
rs2288368 Chr.12: 51299573-51299573 on Build GRCh38
rs758431238 Chr.12: 51299695-51299695 on Build GRCh38
rs576769629 Chr.12: 51299684-51299684 on Build GRCh38
rs748751315 Chr.12: 51299561-51299561 on Build GRCh38
rs761737319 Chr.12: 51299672-51299672 on Build GRCh38
rs746806256 Chr.12: 51299697-51299697 on Build GRCh38
rs1037771525 Chr.12: 51299539-51299539 on Build GRCh38
rs764653272 Chr.12: 51299610-51299610 on Build GRCh38
rs768661735 Chr.12: 51299708-51299708 on Build GRCh38
rs751056621 Chr.12: 51299737-51299738 on Build GRCh38

View Full Product Details