Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79120181-79120410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHMT
Entrez Gene ID
635
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
BHMT1, HEL-S-61p
UniGene
Hs.80756
Gene Chromosome Location
Chr.5: 79111781-79132290 on Build GRCh38
Transcripts
NM_001713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104713 c.337G>A p.E113K Chr.5: 79120401-79120401 on Build GRCh38
3074635 c.312C>T p.C104C Chr.5: 79120376-79120376 on Build GRCh38
3074636 c.322C>T p.R108* Chr.5: 79120386-79120386 on Build GRCh38
259633 c.323G>A p.R108Q Chr.5: 79120387-79120387 on Build GRCh38
5960239 c.292_293delGA p.E98fs*3 Chr.5: 79120356-79120357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375977888 Chr.5: 79120300-79120300 on Build GRCh38
rs371204172 Chr.5: 79120326-79120326 on Build GRCh38
rs1042530000 Chr.5: 79120262-79120262 on Build GRCh38
rs111297335 Chr.5: 79120387-79120387 on Build GRCh38
rs199624450 Chr.5: 79120382-79120382 on Build GRCh38
rs749230412 Chr.5: 79120328-79120328 on Build GRCh38
rs971079538 Chr.5: 79120282-79120282 on Build GRCh38
rs551968640 Chr.5: 79120233-79120233 on Build GRCh38
rs776832617 Chr.5: 79120326-79120327 on Build GRCh38
rs374780042 Chr.5: 79120350-79120350 on Build GRCh38
rs114070281 Chr.5: 79120225-79120225 on Build GRCh38
rs760321372 Chr.5: 79120364-79120364 on Build GRCh38
rs932659680 Chr.5: 79120218-79120218 on Build GRCh38
rs200348644 Chr.5: 79120376-79120376 on Build GRCh38
rs547503968 Chr.5: 79120336-79120336 on Build GRCh38
rs754328429 Chr.5: 79120377-79120377 on Build GRCh38
rs201674272 Chr.5: 79120330-79120330 on Build GRCh38
rs1032726301 Chr.5: 79120311-79120311 on Build GRCh38
rs35617181 Chr.5: 79120386-79120387 on Build GRCh38
rs904064532 Chr.5: 79120292-79120292 on Build GRCh38
rs562583365 Chr.5: 79120255-79120255 on Build GRCh38
rs894122242 Chr.5: 79120383-79120383 on Build GRCh38
rs774227418 Chr.5: 79120332-79120332 on Build GRCh38
rs747129480 Chr.5: 79120305-79120305 on Build GRCh38
rs761333211 Chr.5: 79120338-79120338 on Build GRCh38
rs767239857 Chr.5: 79120340-79120340 on Build GRCh38
rs368232444 Chr.5: 79120312-79120312 on Build GRCh38

View Full Product Details