Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168944450-168944723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB11
Entrez Gene ID
8647
Gene Name
ATP binding cassette subfamily B member 11
Gene Aliases
ABC16, BRIC2, BSEP, PFIC-2, PFIC2, PGY4, SPGP
UniGene
Hs.658439
Gene Chromosome Location
Chr.2: 168915468-169031396 on Build GRCh38
Transcripts
NM_003742.2 XM_006712817.3 XM_011512077.2 XM_011512078.2 XM_011512080.2 XM_011512081.2 XM_017005165.1 XM_017005166.1 XM_017005167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009122 c.2494C>T p.R832C Chr.2: 168944721-168944721 on Build GRCh38
4816802 c.2550C>T p.L850L Chr.2: 168944665-168944665 on Build GRCh38
366342 c.2543A>T p.D848V Chr.2: 168944672-168944672 on Build GRCh38
5465174 c.2547C>T p.D849D Chr.2: 168944668-168944668 on Build GRCh38
1729251 c.2497A>C p.K833Q Chr.2: 168944718-168944718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031723171 Chr.2: 168944502-168944502 on Build GRCh38
rs199760469 Chr.2: 168944570-168944570 on Build GRCh38
rs544968183 Chr.2: 168944517-168944517 on Build GRCh38
rs755766647 Chr.2: 168944645-168944645 on Build GRCh38
rs200829550 Chr.2: 168944597-168944597 on Build GRCh38
rs760922641 Chr.2: 168944623-168944623 on Build GRCh38
rs575962775 Chr.2: 168944498-168944498 on Build GRCh38
rs367874361 Chr.2: 168944692-168944692 on Build GRCh38
rs1014304470 Chr.2: 168944650-168944650 on Build GRCh38
rs774541851 Chr.2: 168944598-168944598 on Build GRCh38
rs754217921 Chr.2: 168944641-168944641 on Build GRCh38
rs562159060 Chr.2: 168944496-168944496 on Build GRCh38
rs943580228 Chr.2: 168944531-168944531 on Build GRCh38
rs868431063 Chr.2: 168944589-168944589 on Build GRCh38
rs1039244570 Chr.2: 168944533-168944533 on Build GRCh38
rs564465398 Chr.2: 168944560-168944560 on Build GRCh38
rs377240498 Chr.2: 168944688-168944688 on Build GRCh38
rs369231927 Chr.2: 168944695-168944695 on Build GRCh38
rs746872311 Chr.2: 168944583-168944583 on Build GRCh38
rs750005514 Chr.2: 168944647-168944647 on Build GRCh38
rs758118928 Chr.2: 168944579-168944579 on Build GRCh38
rs757842499 Chr.2: 168944656-168944656 on Build GRCh38
rs780904250 Chr.2: 168944594-168944594 on Build GRCh38
rs952745163 Chr.2: 168944607-168944607 on Build GRCh38
rs764575275 Chr.2: 168944640-168944640 on Build GRCh38
rs201068438 Chr.2: 168944606-168944606 on Build GRCh38
rs886055064 Chr.2: 168944622-168944622 on Build GRCh38
rs759524655 Chr.2: 168944605-168944605 on Build GRCh38
rs762423892 Chr.2: 168944642-168944642 on Build GRCh38
rs1017527585 Chr.2: 168944566-168944566 on Build GRCh38
rs995512747 Chr.2: 168944577-168944577 on Build GRCh38
rs765638545 Chr.2: 168944646-168944646 on Build GRCh38
rs118109635 Chr.2: 168944621-168944621 on Build GRCh38
rs11568374 Chr.2: 168944587-168944587 on Build GRCh38
rs767098559 Chr.2: 168944511-168944511 on Build GRCh38
rs754572557 Chr.2: 168944694-168944694 on Build GRCh38

View Full Product Details