Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55377834-55378067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
UniGene
Hs.414809
Gene Chromosome Location
Chr.14: 55366391-55411858 on Build GRCh38
Transcripts
NM_014924.4 XM_011536563.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1255619 c.1099G>C p.D367H Chr.14: 55377892-55377892 on Build GRCh38
6606604 c.1082C>A p.S361Y Chr.14: 55377988-55377988 on Build GRCh38
3764498 c.1003T>C p.C335R Chr.14: 55378067-55378067 on Build GRCh38
3769464 c.1036C>T p.R346* Chr.14: 55378034-55378034 on Build GRCh38
5519256 c.1087-4_1087-3insT p.? Chr.14: 55377907-55377908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762727034 Chr.14: 55377887-55377887 on Build GRCh38
rs374651173 Chr.14: 55378033-55378033 on Build GRCh38
rs376183173 Chr.14: 55377963-55377963 on Build GRCh38
rs764746238 Chr.14: 55377920-55377920 on Build GRCh38
rs781368580 Chr.14: 55377999-55377999 on Build GRCh38
rs200769555 Chr.14: 55377898-55377898 on Build GRCh38
rs990781050 Chr.14: 55378001-55378001 on Build GRCh38
rs780668794 Chr.14: 55377906-55377907 on Build GRCh38
rs758872579 Chr.14: 55377859-55377859 on Build GRCh38
rs373428312 Chr.14: 55377856-55377856 on Build GRCh38
rs780490392 Chr.14: 55377870-55377870 on Build GRCh38
rs199841780 Chr.14: 55377880-55377880 on Build GRCh38
rs752226787 Chr.14: 55377933-55377933 on Build GRCh38
rs564212082 Chr.14: 55377971-55377971 on Build GRCh38
rs370639512 Chr.14: 55377916-55377916 on Build GRCh38
rs766886256 Chr.14: 55377962-55377962 on Build GRCh38
rs528347340 Chr.14: 55378025-55378025 on Build GRCh38
rs768903263 Chr.14: 55377873-55377873 on Build GRCh38
rs540139563 Chr.14: 55378034-55378034 on Build GRCh38
rs4898862 Chr.14: 55377993-55377993 on Build GRCh38
rs759016546 Chr.14: 55377917-55377917 on Build GRCh38
rs376657230 Chr.14: 55377877-55377877 on Build GRCh38
rs202211215 Chr.14: 55377882-55377882 on Build GRCh38
rs760263395 Chr.14: 55377936-55377936 on Build GRCh38
rs777918642 Chr.14: 55378028-55378028 on Build GRCh38
rs928379591 Chr.14: 55378042-55378042 on Build GRCh38
rs747317746 Chr.14: 55377973-55377974 on Build GRCh38
rs866182998 Chr.14: 55377878-55377878 on Build GRCh38
rs971318943 Chr.14: 55378009-55378009 on Build GRCh38
rs370308161 Chr.14: 55378013-55378013 on Build GRCh38
rs991095620 Chr.14: 55377891-55377891 on Build GRCh38
rs373243634 Chr.14: 55377949-55377949 on Build GRCh38
rs756359175 Chr.14: 55378021-55378021 on Build GRCh38
rs550659701 Chr.14: 55377940-55377940 on Build GRCh38
rs758746017 Chr.14: 55377957-55377957 on Build GRCh38
rs1030902593 Chr.14: 55377874-55377874 on Build GRCh38
rs772996120 Chr.14: 55377886-55377886 on Build GRCh38
rs747071676 Chr.14: 55377872-55377872 on Build GRCh38

View Full Product Details