Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1010857-1011116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395242 c.1310T>A p.F437Y Chr.19: 1011103-1011103 on Build GRCh38
4073699 c.1348G>A p.E450K Chr.19: 1010865-1010865 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557482759 Chr.19: 1011030-1011030 on Build GRCh38
rs527732986 Chr.19: 1010929-1010929 on Build GRCh38
rs750731111 Chr.19: 1011060-1011060 on Build GRCh38
rs768755088 Chr.19: 1011072-1011072 on Build GRCh38
rs750074352 Chr.19: 1010888-1010888 on Build GRCh38
rs1035797313 Chr.19: 1010973-1010973 on Build GRCh38
rs374335527 Chr.19: 1011083-1011083 on Build GRCh38
rs760299079 Chr.19: 1010895-1010895 on Build GRCh38
rs749325371 Chr.19: 1010919-1010919 on Build GRCh38
rs1029608828 Chr.19: 1011088-1011088 on Build GRCh38
rs370734644 Chr.19: 1010930-1010930 on Build GRCh38
rs796149653 Chr.19: 1010909-1010911 on Build GRCh38
rs561567447 Chr.19: 1010908-1010908 on Build GRCh38
rs759082266 Chr.19: 1010941-1010941 on Build GRCh38
rs554493447 Chr.19: 1010983-1010983 on Build GRCh38
rs373234767 Chr.19: 1010924-1010924 on Build GRCh38
rs780323026 Chr.19: 1011064-1011064 on Build GRCh38
rs757723119 Chr.19: 1011055-1011055 on Build GRCh38
rs1041171338 Chr.19: 1011033-1011033 on Build GRCh38
rs768547126 Chr.19: 1010923-1010923 on Build GRCh38
rs751793796 Chr.19: 1010884-1010884 on Build GRCh38
rs781433078 Chr.19: 1010912-1010912 on Build GRCh38
rs752076857 Chr.19: 1011047-1011047 on Build GRCh38
rs560277415 Chr.19: 1010913-1010913 on Build GRCh38
rs368879268 Chr.19: 1010900-1010900 on Build GRCh38
rs550372795 Chr.19: 1010968-1010968 on Build GRCh38
rs11665815 Chr.19: 1010904-1010904 on Build GRCh38
rs769956423 Chr.19: 1010915-1010915 on Build GRCh38
rs998750968 Chr.19: 1010889-1010889 on Build GRCh38
rs8100391 Chr.19: 1010974-1010974 on Build GRCh38
rs985857877 Chr.19: 1011008-1011008 on Build GRCh38
rs887905112 Chr.19: 1010986-1010986 on Build GRCh38
rs188394433 Chr.19: 1010905-1010905 on Build GRCh38
rs536143795 Chr.19: 1010981-1010981 on Build GRCh38
rs749390275 Chr.19: 1011066-1011066 on Build GRCh38
rs565515644 Chr.19: 1010992-1010992 on Build GRCh38
rs370060644 Chr.19: 1011059-1011059 on Build GRCh38
rs972976620 Chr.19: 1010985-1010985 on Build GRCh38
rs985272470 Chr.19: 1010883-1010883 on Build GRCh38
rs539453218 Chr.19: 1010999-1010999 on Build GRCh38
rs111712574 Chr.19: 1011095-1011095 on Build GRCh38
rs767333411 Chr.19: 1010885-1010885 on Build GRCh38
rs772084120 Chr.19: 1011074-1011074 on Build GRCh38
rs1056656804 Chr.19: 1010958-1010958 on Build GRCh38
rs375122435 Chr.19: 1010943-1010943 on Build GRCh38
rs931623105 Chr.19: 1011006-1011006 on Build GRCh38
rs918147479 Chr.19: 1010962-1010962 on Build GRCh38
rs756337622 Chr.19: 1011061-1011061 on Build GRCh38
rs552369521 Chr.19: 1010938-1010938 on Build GRCh38
rs778753134 Chr.19: 1011073-1011073 on Build GRCh38
rs113349320 Chr.19: 1010948-1010948 on Build GRCh38
rs911256519 Chr.19: 1011029-1011029 on Build GRCh38
rs766014308 Chr.19: 1010899-1010899 on Build GRCh38
rs753290731 Chr.19: 1010940-1010940 on Build GRCh38
rs780124339 Chr.19: 1010917-1010917 on Build GRCh38
rs7248769 Chr.19: 1010909-1010909 on Build GRCh38
rs377631297 Chr.19: 1010926-1010926 on Build GRCh38

View Full Product Details