Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120739363-120739598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADS
Entrez Gene ID
35
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Gene Aliases
ACAD3, SCAD
UniGene
Hs.507076
Gene Chromosome Location
Chr.12: 120725738-120740008 on Build GRCh38
Transcripts
NM_000017.3 NM_001302554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238654 c.1156C>T p.R386C Chr.12: 120739365-120739365 on Build GRCh38
5840325 c.1211G>A p.G404E Chr.12: 120739420-120739420 on Build GRCh38
6591692 c.1231C>T p.R411W Chr.12: 120739440-120739440 on Build GRCh38
39701 c.1200G>A p.L400L Chr.12: 120739409-120739409 on Build GRCh38
1299010 c.1186G>A p.E396K Chr.12: 120739395-120739395 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760829554 Chr.12: 120739455-120739455 on Build GRCh38
rs895841347 Chr.12: 120739385-120739385 on Build GRCh38
rs775999272 Chr.12: 120739391-120739391 on Build GRCh38
rs374043146 Chr.12: 120739415-120739415 on Build GRCh38
rs775747840 Chr.12: 120739454-120739454 on Build GRCh38
rs780588449 Chr.12: 120739483-120739483 on Build GRCh38
rs372567027 Chr.12: 120739484-120739484 on Build GRCh38
rs779465933 Chr.12: 120739486-120739486 on Build GRCh38
rs754619218 Chr.12: 120739476-120739476 on Build GRCh38
rs369840561 Chr.12: 120739407-120739407 on Build GRCh38
rs2229533 Chr.12: 120739453-120739453 on Build GRCh38
rs375931905 Chr.12: 120739404-120739404 on Build GRCh38
rs926187308 Chr.12: 120739405-120739405 on Build GRCh38
rs532956280 Chr.12: 120739538-120739538 on Build GRCh38
rs757155952 Chr.12: 120739411-120739411 on Build GRCh38
rs771419406 Chr.12: 120739441-120739441 on Build GRCh38
rs774815274 Chr.12: 120739452-120739452 on Build GRCh38
rs866546417 Chr.12: 120739410-120739410 on Build GRCh38
rs778608148 Chr.12: 120739412-120739412 on Build GRCh38
rs761682729 Chr.12: 120739458-120739458 on Build GRCh38
rs377069922 Chr.12: 120739418-120739418 on Build GRCh38
rs772439987 Chr.12: 120739490-120739490 on Build GRCh38
rs948685149 Chr.12: 120739392-120739392 on Build GRCh38
rs545737847 Chr.12: 120739419-120739419 on Build GRCh38
rs377080695 Chr.12: 120739485-120739485 on Build GRCh38
rs759774887 Chr.12: 120739394-120739394 on Build GRCh38
rs556890619 Chr.12: 120739416-120739416 on Build GRCh38
rs916490617 Chr.12: 120739520-120739520 on Build GRCh38
rs754897003 Chr.12: 120739457-120739457 on Build GRCh38
rs369416846 Chr.12: 120739440-120739440 on Build GRCh38
rs926065750 Chr.12: 120739577-120739577 on Build GRCh38
rs561649047 Chr.12: 120739513-120739513 on Build GRCh38
rs967435328 Chr.12: 120739535-120739535 on Build GRCh38
rs2229534 Chr.12: 120739563-120739563 on Build GRCh38
rs886049031 Chr.12: 120739539-120739539 on Build GRCh38
rs765271596 Chr.12: 120739464-120739464 on Build GRCh38
rs1009003917 Chr.12: 120739533-120739533 on Build GRCh38
rs767774362 Chr.12: 120739401-120739401 on Build GRCh38
rs749790770 Chr.12: 120739435-120739435 on Build GRCh38
rs374762327 Chr.12: 120739534-120739534 on Build GRCh38
rs776136316 Chr.12: 120739422-120739422 on Build GRCh38
rs779933328 Chr.12: 120739474-120739474 on Build GRCh38
rs746361724 Chr.12: 120739487-120739487 on Build GRCh38
rs371172924 Chr.12: 120739529-120739529 on Build GRCh38
rs188190117 Chr.12: 120739475-120739475 on Build GRCh38
rs900872014 Chr.12: 120739527-120739527 on Build GRCh38
rs775765818 Chr.12: 120739495-120739495 on Build GRCh38
rs1033308144 Chr.12: 120739558-120739558 on Build GRCh38
rs758346545 Chr.12: 120739485-120739486 on Build GRCh38
rs3916 Chr.12: 120739469-120739469 on Build GRCh38
rs992102695 Chr.12: 120739506-120739506 on Build GRCh38

View Full Product Details