Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138162269-138162494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBR1
Entrez Gene ID
51163
Gene Name
debranching RNA lariats 1
Gene Aliases
-
UniGene
Hs.477700
Gene Chromosome Location
Chr.3: 138160988-138174949 on Build GRCh38
Transcripts
NM_016216.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6902746 c.1053C>A p.S351R Chr.3: 138162471-138162471 on Build GRCh38
1616971 c.1199A>G p.D400G Chr.3: 138162325-138162325 on Build GRCh38
3695838 c.1062G>T p.Q354H Chr.3: 138162462-138162462 on Build GRCh38
7269036 c.1044T>C p.Y348Y Chr.3: 138162480-138162480 on Build GRCh38
7259729 c.1116C>A p.A372A Chr.3: 138162408-138162408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754271435 Chr.3: 138162412-138162412 on Build GRCh38
rs200452995 Chr.3: 138162321-138162321 on Build GRCh38
rs866987440 Chr.3: 138162428-138162428 on Build GRCh38
rs766308614 Chr.3: 138162341-138162341 on Build GRCh38
rs746423866 Chr.3: 138162432-138162432 on Build GRCh38
rs931139638 Chr.3: 138162442-138162442 on Build GRCh38
rs772853801 Chr.3: 138162467-138162467 on Build GRCh38
rs972657614 Chr.3: 138162427-138162427 on Build GRCh38
rs752794375 Chr.3: 138162385-138162385 on Build GRCh38
rs3732839 Chr.3: 138162423-138162423 on Build GRCh38
rs893968481 Chr.3: 138162305-138162305 on Build GRCh38
rs769445912 Chr.3: 138162320-138162320 on Build GRCh38
rs373179901 Chr.3: 138162441-138162441 on Build GRCh38
rs1011366400 Chr.3: 138162318-138162318 on Build GRCh38
rs762590832 Chr.3: 138162471-138162471 on Build GRCh38
rs149959691 Chr.3: 138162311-138162311 on Build GRCh38
rs117574050 Chr.3: 138162433-138162433 on Build GRCh38
rs767544832 Chr.3: 138162356-138162356 on Build GRCh38
rs1019540958 Chr.3: 138162401-138162401 on Build GRCh38
rs757760178 Chr.3: 138162419-138162419 on Build GRCh38
rs759457997 Chr.3: 138162355-138162355 on Build GRCh38
rs769344353 Chr.3: 138162449-138162449 on Build GRCh38
rs986839647 Chr.3: 138162398-138162398 on Build GRCh38
rs778007641 Chr.3: 138162410-138162410 on Build GRCh38
rs1028075862 Chr.3: 138162336-138162336 on Build GRCh38
rs201996920 Chr.3: 138162394-138162394 on Build GRCh38
rs144975754 Chr.3: 138162314-138162314 on Build GRCh38
rs369882903 Chr.3: 138162352-138162352 on Build GRCh38
rs747642808 Chr.3: 138162443-138162443 on Build GRCh38
rs768335561 Chr.3: 138162302-138162302 on Build GRCh38

View Full Product Details