Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138152389-138152618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_011543109.1 XM_011543110.1 XM_017008969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365844 c.2742A>G p.E914E Chr.5: 138152596-138152596 on Build GRCh38
3393154 c.2767G>T p.E923* Chr.5: 138152571-138152571 on Build GRCh38
3850368 c.2854G>A p.E952K Chr.5: 138152484-138152484 on Build GRCh38
4666125 c.2751G>A p.P917P Chr.5: 138152587-138152587 on Build GRCh38
5867348 c.2758_2759delGA p.E920fs*2 Chr.5: 138152579-138152580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761633285 Chr.5: 138152469-138152469 on Build GRCh38
rs758983450 Chr.5: 138152522-138152522 on Build GRCh38
rs774316641 Chr.5: 138152570-138152570 on Build GRCh38
rs754201802 Chr.5: 138152467-138152468 on Build GRCh38
rs150159308 Chr.5: 138152558-138152558 on Build GRCh38
rs753328688 Chr.5: 138152504-138152504 on Build GRCh38
rs749230187 Chr.5: 138152457-138152457 on Build GRCh38
rs757679501 Chr.5: 138152533-138152533 on Build GRCh38
rs557433008 Chr.5: 138152455-138152455 on Build GRCh38
rs369431228 Chr.5: 138152542-138152542 on Build GRCh38
rs781386566 Chr.5: 138152536-138152536 on Build GRCh38
rs999897147 Chr.5: 138152587-138152587 on Build GRCh38
rs375407999 Chr.5: 138152490-138152490 on Build GRCh38
rs774155127 Chr.5: 138152468-138152468 on Build GRCh38
rs767127696 Chr.5: 138152527-138152527 on Build GRCh38
rs758978639 Chr.5: 138152488-138152488 on Build GRCh38
rs140870898 Chr.5: 138152557-138152557 on Build GRCh38
rs941544177 Chr.5: 138152526-138152526 on Build GRCh38
rs111670295 Chr.5: 138152444-138152444 on Build GRCh38
rs374460094 Chr.5: 138152588-138152588 on Build GRCh38
rs201588407 Chr.5: 138152471-138152471 on Build GRCh38
rs947720420 Chr.5: 138152544-138152544 on Build GRCh38
rs768523710 Chr.5: 138152464-138152464 on Build GRCh38
rs748015811 Chr.5: 138152577-138152577 on Build GRCh38
rs539478006 Chr.5: 138152453-138152453 on Build GRCh38
rs200412361 Chr.5: 138152560-138152560 on Build GRCh38
rs752319711 Chr.5: 138152528-138152528 on Build GRCh38
rs760376365 Chr.5: 138152597-138152597 on Build GRCh38
rs751136954 Chr.5: 138152481-138152481 on Build GRCh38
rs199793838 Chr.5: 138152591-138152591 on Build GRCh38
rs766051001 Chr.5: 138152496-138152496 on Build GRCh38

View Full Product Details