Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 240093352-240093577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
UniGene
Hs.24889
Gene Chromosome Location
Chr.1: 240013776-240475189 on Build GRCh38
Transcripts
NM_001305424.1 NM_020066.4 XM_011544237.2 XM_017001837.1 XM_017001838.1 XM_017001839.1 XM_017001842.1 XM_017001843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639763 c.1883G>T p.W628L Chr.1: 240093563-240093563 on Build GRCh38
3485953 c.1887G>A p.T629T Chr.1: 240093567-240093567 on Build GRCh38
4143555 c.1842C>A p.A614A Chr.1: 240093522-240093522 on Build GRCh38
3360765 c.1765C>T p.R589W Chr.1: 240093445-240093445 on Build GRCh38
6214311 c.1825G>C p.A609P Chr.1: 240093505-240093505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141492081 Chr.1: 240093475-240093475 on Build GRCh38
rs749834072 Chr.1: 240093551-240093551 on Build GRCh38
rs377516551 Chr.1: 240093459-240093459 on Build GRCh38
rs760197257 Chr.1: 240093429-240093429 on Build GRCh38
rs541648891 Chr.1: 240093549-240093549 on Build GRCh38
rs766197562 Chr.1: 240093520-240093520 on Build GRCh38
rs764285041 Chr.1: 240093469-240093469 on Build GRCh38
rs760382584 Chr.1: 240093498-240093498 on Build GRCh38
rs776179455 Chr.1: 240093503-240093503 on Build GRCh38
rs147961923 Chr.1: 240093467-240093467 on Build GRCh38
rs761311789 Chr.1: 240093504-240093504 on Build GRCh38
rs771349783 Chr.1: 240093489-240093489 on Build GRCh38
rs973142541 Chr.1: 240093458-240093458 on Build GRCh38
rs374512507 Chr.1: 240093372-240093372 on Build GRCh38
rs546535429 Chr.1: 240093439-240093439 on Build GRCh38
rs201430864 Chr.1: 240093398-240093398 on Build GRCh38
rs142335257 Chr.1: 240093481-240093481 on Build GRCh38
rs867051143 Chr.1: 240093392-240093392 on Build GRCh38
rs568813967 Chr.1: 240093471-240093471 on Build GRCh38
rs920230810 Chr.1: 240093389-240093389 on Build GRCh38
rs756242155 Chr.1: 240093542-240093542 on Build GRCh38
rs10926124 Chr.1: 240093522-240093522 on Build GRCh38
rs768799931 Chr.1: 240093378-240093378 on Build GRCh38
rs773768122 Chr.1: 240093402-240093402 on Build GRCh38
rs781434131 Chr.1: 240093477-240093477 on Build GRCh38
rs571454130 Chr.1: 240093441-240093441 on Build GRCh38
rs201558855 Chr.1: 240093518-240093518 on Build GRCh38
rs150033699 Chr.1: 240093393-240093393 on Build GRCh38
rs368925775 Chr.1: 240093403-240093403 on Build GRCh38
rs377670694 Chr.1: 240093554-240093554 on Build GRCh38
rs1013760774 Chr.1: 240093384-240093384 on Build GRCh38
rs727502860 Chr.1: 240093503-240093504 on Build GRCh38
rs145379416 Chr.1: 240093461-240093461 on Build GRCh38
rs748222847 Chr.1: 240093479-240093479 on Build GRCh38
rs747283262 Chr.1: 240093473-240093473 on Build GRCh38
rs374209120 Chr.1: 240093537-240093537 on Build GRCh38
rs949474137 Chr.1: 240093470-240093470 on Build GRCh38
rs762413223 Chr.1: 240093394-240093394 on Build GRCh38
rs752375308 Chr.1: 240093415-240093415 on Build GRCh38
rs767117704 Chr.1: 240093414-240093414 on Build GRCh38
rs528260990 Chr.1: 240093371-240093371 on Build GRCh38
rs138052423 Chr.1: 240093486-240093486 on Build GRCh38
rs774540174 Chr.1: 240093491-240093491 on Build GRCh38
rs755308785 Chr.1: 240093526-240093526 on Build GRCh38
rs554698558 Chr.1: 240093499-240093499 on Build GRCh38
rs763369839 Chr.1: 240093407-240093407 on Build GRCh38
rs750394019 Chr.1: 240093519-240093519 on Build GRCh38
rs997168836 Chr.1: 240093517-240093517 on Build GRCh38
rs770488060 Chr.1: 240093480-240093480 on Build GRCh38

View Full Product Details