Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75764785-75765030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALK1
Entrez Gene ID
2584
Gene Name
galactokinase 1
Gene Aliases
GALK, GK1, HEL-S-19
UniGene
Hs.407966
Gene Chromosome Location
Chr.17: 75757937-75765199 on Build GRCh38
Transcripts
NM_000154.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4648088 c.155T>A p.V52E Chr.17: 75764982-75764982 on Build GRCh38
4922996 c.163A>G p.M55V Chr.17: 75764974-75764974 on Build GRCh38
6302696 c.145C>A p.Q49K Chr.17: 75764992-75764992 on Build GRCh38
6036544 c.165+5G>A p.? Chr.17: 75764967-75764967 on Build GRCh38
4527789 c.148G>A p.G50S Chr.17: 75764989-75764989 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs914086622 Chr.17: 75764924-75764924 on Build GRCh38
rs553868612 Chr.17: 75764897-75764897 on Build GRCh38
rs771513393 Chr.17: 75764822-75764822 on Build GRCh38
rs574322744 Chr.17: 75764889-75764890 on Build GRCh38
rs775058415 Chr.17: 75764861-75764861 on Build GRCh38
rs377565047 Chr.17: 75764809-75764810 on Build GRCh38
rs749705043 Chr.17: 75764816-75764816 on Build GRCh38
rs969718018 Chr.17: 75764839-75764839 on Build GRCh38
rs995270898 Chr.17: 75764814-75764814 on Build GRCh38
rs376599506 Chr.17: 75765003-75765003 on Build GRCh38
rs760408680 Chr.17: 75764935-75764935 on Build GRCh38
rs1001190932 Chr.17: 75764865-75764865 on Build GRCh38
rs750227939 Chr.17: 75764959-75764959 on Build GRCh38
rs978533427 Chr.17: 75764886-75764887 on Build GRCh38
rs746381829 Chr.17: 75764993-75764993 on Build GRCh38
rs925336919 Chr.17: 75764974-75764974 on Build GRCh38
rs534012909 Chr.17: 75764887-75764887 on Build GRCh38
rs774696699 Chr.17: 75764828-75764828 on Build GRCh38
rs573790143 Chr.17: 75764984-75764984 on Build GRCh38
rs1026693481 Chr.17: 75764818-75764818 on Build GRCh38
rs763464166 Chr.17: 75764948-75764948 on Build GRCh38
rs767885167 Chr.17: 75764840-75764840 on Build GRCh38
rs375582200 Chr.17: 75764967-75764967 on Build GRCh38
rs761443332 Chr.17: 75765014-75765014 on Build GRCh38
rs765331402 Chr.17: 75764951-75764951 on Build GRCh38
rs370714053 Chr.17: 75765013-75765013 on Build GRCh38
rs758207969 Chr.17: 75764965-75764965 on Build GRCh38
rs775856143 Chr.17: 75764998-75764998 on Build GRCh38
rs960969163 Chr.17: 75764890-75764890 on Build GRCh38
rs756377738 Chr.17: 75764971-75764971 on Build GRCh38
rs749455293 Chr.17: 75764981-75764981 on Build GRCh38
rs779996721 Chr.17: 75764966-75764966 on Build GRCh38
rs760223863 Chr.17: 75765001-75765001 on Build GRCh38
rs146045315 Chr.17: 75764976-75764976 on Build GRCh38
rs966826174 Chr.17: 75764947-75764947 on Build GRCh38
rs779337940 Chr.17: 75764991-75764991 on Build GRCh38
rs992764445 Chr.17: 75764909-75764909 on Build GRCh38
rs772388628 Chr.17: 75764994-75764994 on Build GRCh38
rs1034384456 Chr.17: 75764889-75764889 on Build GRCh38
rs761623771 Chr.17: 75764949-75764949 on Build GRCh38

View Full Product Details