Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186646163-186646436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FETUB
Entrez Gene ID
26998
Gene Name
fetuin B
Gene Aliases
16G2, Gugu, IRL685
UniGene
Hs.81073
Gene Chromosome Location
Chr.3: 186635828-186653008 on Build GRCh38
Transcripts
NM_001308077.1 NM_001308079.1 NM_014375.2 XM_005247351.3 XM_011512681.2 XM_011512682.2 XM_011512683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042322 c.637G>T p.E213* Chr.3: 186646290-186646290 on Build GRCh38
4827103 c.641C>T p.S214L Chr.3: 186646294-186646294 on Build GRCh38
5795378 c.691T>A p.S231T Chr.3: 186646344-186646344 on Build GRCh38
479822 c.675A>T p.S225S Chr.3: 186646328-186646328 on Build GRCh38
4115952 c.656C>T p.S219F Chr.3: 186646309-186646309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762685404 Chr.3: 186646379-186646379 on Build GRCh38
rs73886004 Chr.3: 186646280-186646280 on Build GRCh38
rs139111101 Chr.3: 186646260-186646260 on Build GRCh38
rs778018393 Chr.3: 186646203-186646203 on Build GRCh38
rs759546734 Chr.3: 186646279-186646279 on Build GRCh38
rs532837954 Chr.3: 186646192-186646192 on Build GRCh38
rs761248477 Chr.3: 186646253-186646253 on Build GRCh38
rs150905870 Chr.3: 186646281-186646281 on Build GRCh38
rs747298078 Chr.3: 186646337-186646337 on Build GRCh38
rs376462332 Chr.3: 186646275-186646275 on Build GRCh38
rs757555767 Chr.3: 186646228-186646228 on Build GRCh38
rs746274339 Chr.3: 186646236-186646236 on Build GRCh38
rs551036685 Chr.3: 186646373-186646373 on Build GRCh38
rs371042019 Chr.3: 186646302-186646302 on Build GRCh38
rs529475246 Chr.3: 186646341-186646341 on Build GRCh38
rs749337242 Chr.3: 186646207-186646207 on Build GRCh38
rs372437277 Chr.3: 186646238-186646238 on Build GRCh38
rs772479748 Chr.3: 186646237-186646237 on Build GRCh38
rs189137781 Chr.3: 186646287-186646287 on Build GRCh38
rs866128504 Chr.3: 186646393-186646393 on Build GRCh38
rs199763365 Chr.3: 186646340-186646340 on Build GRCh38
rs770447808 Chr.3: 186646382-186646382 on Build GRCh38
rs747718049 Chr.3: 186646365-186646365 on Build GRCh38
rs368526622 Chr.3: 186646376-186646376 on Build GRCh38
rs758739763 Chr.3: 186646310-186646310 on Build GRCh38
rs538768106 Chr.3: 186646316-186646316 on Build GRCh38
rs776150518 Chr.3: 186646248-186646248 on Build GRCh38
rs769951694 Chr.3: 186646253-186646253 on Build GRCh38
rs764885301 Chr.3: 186646256-186646256 on Build GRCh38
rs180922291 Chr.3: 186646377-186646377 on Build GRCh38
rs868842102 Chr.3: 186646240-186646240 on Build GRCh38
rs1045153647 Chr.3: 186646191-186646191 on Build GRCh38
rs6785067 Chr.3: 186646257-186646257 on Build GRCh38
rs766170825 Chr.3: 186646262-186646262 on Build GRCh38
rs769103905 Chr.3: 186646242-186646242 on Build GRCh38
rs905278659 Chr.3: 186646211-186646211 on Build GRCh38
rs139353499 Chr.3: 186646299-186646299 on Build GRCh38
rs756192361 Chr.3: 186646202-186646202 on Build GRCh38
rs751514910 Chr.3: 186646199-186646199 on Build GRCh38
rs370744934 Chr.3: 186646217-186646217 on Build GRCh38
rs964140776 Chr.3: 186646300-186646300 on Build GRCh38
rs751259988 Chr.3: 186646272-186646272 on Build GRCh38
rs200656071 Chr.3: 186646232-186646232 on Build GRCh38
rs747528344 Chr.3: 186646241-186646241 on Build GRCh38
rs892839203 Chr.3: 186646348-186646348 on Build GRCh38

View Full Product Details