Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 130145090-130145361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX3
Entrez Gene ID
10539
Gene Name
glutaredoxin 3
Gene Aliases
GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3
UniGene
Hs.42644
Gene Chromosome Location
Chr.10: 130136375-130182877 on Build GRCh38
Transcripts
NM_001199868.1 NM_001321980.1 NM_006541.4 XM_017015474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4413275 c.100C>A p.L34I Chr.10: 130145218-130145218 on Build GRCh38
3382746 c.94T>G p.S32A Chr.10: 130145212-130145212 on Build GRCh38
3382744 c.93-2A>C p.? Chr.10: 130145209-130145209 on Build GRCh38
916200 c.175G>T p.E59* Chr.10: 130145293-130145293 on Build GRCh38
398869 c.163G>C p.E55Q Chr.10: 130145281-130145281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573814328 Chr.10: 130145229-130145229 on Build GRCh38
rs7073590 Chr.10: 130145159-130145159 on Build GRCh38
rs750613292 Chr.10: 130145276-130145276 on Build GRCh38
rs376653137 Chr.10: 130145192-130145192 on Build GRCh38
rs988601658 Chr.10: 130145208-130145208 on Build GRCh38
rs1000931004 Chr.10: 130145175-130145175 on Build GRCh38
rs538063780 Chr.10: 130145239-130145239 on Build GRCh38
rs184021589 Chr.10: 130145222-130145222 on Build GRCh38
rs965815077 Chr.10: 130145238-130145238 on Build GRCh38
rs142769954 Chr.10: 130145274-130145274 on Build GRCh38
rs777514839 Chr.10: 130145237-130145237 on Build GRCh38
rs769835521 Chr.10: 130145268-130145268 on Build GRCh38
rs188360612 Chr.10: 130145269-130145269 on Build GRCh38
rs760938061 Chr.10: 130145278-130145278 on Build GRCh38
rs754271680 Chr.10: 130145283-130145283 on Build GRCh38
rs759377537 Chr.10: 130145253-130145253 on Build GRCh38
rs560391020 Chr.10: 130145242-130145242 on Build GRCh38
rs7095590 Chr.10: 130145120-130145120 on Build GRCh38
rs770816758 Chr.10: 130145241-130145241 on Build GRCh38
rs895305711 Chr.10: 130145195-130145195 on Build GRCh38
rs753162172 Chr.10: 130145314-130145314 on Build GRCh38
rs201414830 Chr.10: 130145299-130145299 on Build GRCh38
rs370009897 Chr.10: 130145244-130145244 on Build GRCh38
rs200546213 Chr.10: 130145318-130145318 on Build GRCh38
rs1001437961 Chr.10: 130145128-130145128 on Build GRCh38
rs766719697 Chr.10: 130145279-130145279 on Build GRCh38
rs372375546 Chr.10: 130145255-130145255 on Build GRCh38
rs775246586 Chr.10: 130145271-130145271 on Build GRCh38
rs146076334 Chr.10: 130145296-130145296 on Build GRCh38
rs775375212 Chr.10: 130145253-130145254 on Build GRCh38
rs779422263 Chr.10: 130145300-130145300 on Build GRCh38
rs11540092 Chr.10: 130145249-130145249 on Build GRCh38
rs560791157 Chr.10: 130145138-130145138 on Build GRCh38
rs933312279 Chr.10: 130145124-130145124 on Build GRCh38
rs762504680 Chr.10: 130145287-130145287 on Build GRCh38

View Full Product Details