Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55081922-55082175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L1
Entrez Gene ID
54869
Gene Name
EPS8 like 1
Gene Aliases
DRC3, EPS8R1, PP10566
UniGene
Hs.438862
Gene Chromosome Location
Chr.19: 55075853-55087923 on Build GRCh38
Transcripts
NM_017729.3 NM_133180.2 XM_005259020.1 XM_011527050.1 XM_011527051.2 XM_011527052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681454 c.936G>A p.S312S Chr.19: 55082126-55082126 on Build GRCh38
1396486 c.571G>A p.D191N Chr.19: 55082142-55082142 on Build GRCh38
4681456 c.882G>A p.S294S Chr.19: 55082126-55082126 on Build GRCh38
5123145 c.865C>T p.R289W Chr.19: 55082109-55082109 on Build GRCh38
4681455 c.555G>A p.S185S Chr.19: 55082126-55082126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752633580 Chr.19: 55082010-55082010 on Build GRCh38
rs866334640 Chr.19: 55082118-55082118 on Build GRCh38
rs751356074 Chr.19: 55082125-55082125 on Build GRCh38
rs2290474 Chr.19: 55082073-55082073 on Build GRCh38
rs769306289 Chr.19: 55082114-55082114 on Build GRCh38
rs952838328 Chr.19: 55082110-55082110 on Build GRCh38
rs1057009239 Chr.19: 55082028-55082028 on Build GRCh38
rs776055088 Chr.19: 55081995-55081995 on Build GRCh38
rs757456050 Chr.19: 55082040-55082040 on Build GRCh38
rs1000469249 Chr.19: 55082095-55082095 on Build GRCh38
rs148596546 Chr.19: 55082121-55082121 on Build GRCh38
rs751801644 Chr.19: 55082039-55082039 on Build GRCh38
rs373200328 Chr.19: 55082140-55082140 on Build GRCh38
rs754369542 Chr.19: 55082147-55082147 on Build GRCh38
rs1044811187 Chr.19: 55082063-55082063 on Build GRCh38
rs763840536 Chr.19: 55082122-55082122 on Build GRCh38
rs776379223 Chr.19: 55082100-55082100 on Build GRCh38
rs369451381 Chr.19: 55082136-55082136 on Build GRCh38
rs765662256 Chr.19: 55082042-55082042 on Build GRCh38
rs765121339 Chr.19: 55082007-55082007 on Build GRCh38
rs749871929 Chr.19: 55082074-55082074 on Build GRCh38
rs780350918 Chr.19: 55082144-55082144 on Build GRCh38
rs763934234 Chr.19: 55082018-55082018 on Build GRCh38
rs756314632 Chr.19: 55081988-55081996 on Build GRCh38
rs377304711 Chr.19: 55082097-55082097 on Build GRCh38
rs529676368 Chr.19: 55082059-55082059 on Build GRCh38
rs758129876 Chr.19: 55082015-55082015 on Build GRCh38
rs760315869 Chr.19: 55081990-55081990 on Build GRCh38
rs569003413 Chr.19: 55082145-55082145 on Build GRCh38
rs779185278 Chr.19: 55082077-55082077 on Build GRCh38
rs776160232 Chr.19: 55081945-55081945 on Build GRCh38
rs903068933 Chr.19: 55082050-55082050 on Build GRCh38
rs1027194723 Chr.19: 55082106-55082106 on Build GRCh38
rs750674199 Chr.19: 55082131-55082131 on Build GRCh38
rs770249866 Chr.19: 55081944-55081944 on Build GRCh38
rs976751072 Chr.19: 55081956-55081956 on Build GRCh38
rs759109095 Chr.19: 55082003-55082003 on Build GRCh38
rs552338158 Chr.19: 55082130-55082130 on Build GRCh38
rs761688226 Chr.19: 55082126-55082126 on Build GRCh38
rs921368972 Chr.19: 55081958-55081958 on Build GRCh38
rs538588765 Chr.19: 55082104-55082104 on Build GRCh38
rs921241920 Chr.19: 55082143-55082143 on Build GRCh38
rs758902291 Chr.19: 55082113-55082113 on Build GRCh38
rs765881437 Chr.19: 55081994-55081994 on Build GRCh38
rs17700138 Chr.19: 55082049-55082049 on Build GRCh38
rs769142263 Chr.19: 55082076-55082076 on Build GRCh38
rs778795158 Chr.19: 55082152-55082154 on Build GRCh38
rs756291222 Chr.19: 55082056-55082056 on Build GRCh38
rs977161548 Chr.19: 55082133-55082133 on Build GRCh38
rs550451318 Chr.19: 55081947-55081947 on Build GRCh38
rs769158966 Chr.19: 55081981-55081981 on Build GRCh38
rs746653074 Chr.19: 55082084-55082084 on Build GRCh38
rs781305888 Chr.19: 55082041-55082041 on Build GRCh38
rs1019834053 Chr.19: 55082139-55082139 on Build GRCh38
rs896653861 Chr.19: 55082036-55082036 on Build GRCh38
rs772988979 Chr.19: 55081982-55081982 on Build GRCh38

View Full Product Details